Skip to Content
Merck
All Photos(1)

Key Documents

554325

Sigma-Aldrich

Resveratrol

≥98% (HPLC), solid, COX-1 inhibitor, Calbiochem®

Synonym(s):

Resveratrol, trans-3,4ʹ,5-Trihydroxystilbene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O3
CAS Number:
Molecular Weight:
228.24
MDL number:
UNSPSC Code:
12191503
NACRES:
NA.77

Product Name

Resveratrol, A phenolic product found in both grape skins and wines.

Quality Level

description

Merck USA index - 14, 8158

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Oc1cc(cc(c1)\C=C\c2ccc(cc2)O)O

InChI

1S/C14H12O3/c15-12-5-3-10(4-6-12)1-2-11-7-13(16)9-14(17)8-11/h1-9,15-17H/b2-1+

InChI key

LUKBXSAWLPMMSZ-OWOJBTEDSA-N

General description

A phenolic product found in both grape skin and wines. Has anti-fungal, anti-tumor and antioxidative properties. A specific inhibitor of cyclooxygenase-1 (COX-1; ED50 = 15 µM). Also inhibits the hydroxyperoxidase activity of COX-1 (ED50 = 3.7 µM). Inhibits phorbol ester-induced free radical formation in HL-60 cells (ED50 = 27 µM) and acts as an anti-mutagenic agent (ED50 = 15 µM) in TM677 cells treated with DMBA. Also selectively inhibits P450 1A1. Reported to activate sirtuins and promote the survival of eukaryotic cells.
A phenolic product found in both grape skins and wines. Has antifungal, antitumor, and antioxidative properties. A specific inhibitor of cyclooxygenase-1 (COX-1; ED50 = 15 µM). Also inhibits the hydroxyperoxidase activity of COX-1 (ED50 = 3.7 µM). Strongly inhibits insoluble transthyretin (TTR) amyloid fibril formation. Inhibits phorbol ester-induced free radical formation in HL-60 cells (ED50 = 27 µM) and acts as an anti-mutagenic agent (ED50 = 15 µM) in TM677 cells treated with DMBA. Also a selective inhibitor of P450 1A1. Reported to activate sirtuins and promote the survival of eukaryotic cells.

Biochem/physiol Actions

Cell permeable: no
ED50 = 15 µM against COX-1
Primary Target
COX-1
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Howitz, K.T., et al. 2003. Nature425, 191.
Lin, H.Y., et al. 2002. J. Urol.168, 748.
Klabunde, T., et al. 2000. Nat. Struct. Biol.7, 312.
Chun, Y.J., et al. 1999. Biochem. Biophys. Res. Commun.262, 20.
Jang, M., et al. 1997. Science 275, 218.
Chen, C.K., et al. 1996. Gen. Pharmacol.27, 363.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>392.0 °F - closed cup - (External MSDS)

Flash Point(C)

> 200.0 °C - closed cup - (External MSDS)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun Ji Lee et al.
Antioxidants (Basel, Switzerland), 11(6) (2022-06-25)
Emerging evidence shows that peroxisome proliferator-activated receptor delta (PPARδ) plays a pivotal role in cellular aging. However, its function in retinal disease processes such as hyperglycemia-associated diabetic retinopathy is unclear. Here, we demonstrate that PPARδ inhibits premature senescence of retinal
Seungwon An et al.
Cells, 13(5) (2024-03-13)
Chlorine (Cl2) exposure poses a significant risk to ocular health, with the cornea being particularly susceptible to its corrosive effects. Antioxidants, known for their ability to neutralize reactive oxygen species (ROS) and alleviate oxidative stress, were explored as potential therapeutic
Elisa Franzi et al.
International journal of molecular sciences, 24(20) (2023-10-28)
Influenza virus is one of the main causes of respiratory infections worldwide. Despite the availability of seasonal vaccines and antivirals, influenza virus infections cause an important health and economic burden. Therefore, the need to identify alternative antiviral strategies persists. In
Oluwamolakun Bankole et al.
International journal of molecular sciences, 23(3) (2022-02-16)
Amyotrophic lateral sclerosis (ALS) is a fatal adult-onset neurodegenerative disorder. There is no cure and current treatments fail to slow the progression of the disease. Epigenetic modulation in the acetylation state of NF-kB RelA and the histone 3 (H3) protein
Paulina Borkowska et al.
Life (Basel, Switzerland), 12(9) (2022-09-24)
The main problem with using MSC (mesenchymal stem cells) to treat the deficient diseases of the central nervous system is the low cell survival rate after the transplant procedure and their low ability to spontaneously differentiate into functional neurons. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service