Skip to Content
Merck
All Photos(1)

Key Documents

SML3269

Sigma-Aldrich

Losartan potassium

≥98% (HPLC)

Synonym(s):

2-Butyl-4-chloro-1-{[2′-(1H-tetrazol-5-yl)(1,1′-biphenyl)-4-yl]methyl}-1H-imidazole-5-methanol monopotassium salt, 2-butyl-4-chloro-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]- 1H-Imidazole-5-methanol potassium salt, DuP 753, MK 954, Potassium 5-(4′-((2-butyl-4-chloro-5-(hydroxymethyl)-1H-imidazol-1-yl)methyl)-[1,1′-biphenyl]-2-yl)tetrazol-1-ide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22ClKN6O
CAS Number:
Molecular Weight:
461.00
Beilstein:
5845770
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCCCc1nc(Cl)c(CO)n1Cc2ccc(cc2)-c3ccccc3-c4nnnn4[K]

InChI

1S/C22H22ClN6O.K/c1-2-3-8-20-24-21(23)19(14-30)29(20)13-15-9-11-16(12-10-15)17-6-4-5-7-18(17)22-25-27-28-26-22;/h4-7,9-12,30H,2-3,8,13-14H2,1H3;/q-1;+1

InChI key

OXCMYAYHXIHQOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Losartan potassium is an orally available, potent, and selective competitive angiotensin II receptor type 1 (AT1) antagonist. Losartan potassium is used for treatment of hypertension.
orally available, potent, and selective competitive angiotensin II receptor type 1 (AT1) antagonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel P Regan et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 28(4), 662-676 (2021-09-29)
There is increasing recognition that progress in immuno-oncology could be accelerated by evaluating immune-based therapies in dogs with spontaneous cancers. Osteosarcoma (OS) is one tumor for which limited clinical benefit has been observed with the use of immune checkpoint inhibitors.
Yoon-A Park et al.
Journal of personalized medicine, 11(7) (2021-07-03)
This study aimed to investigate the influence of CYP2C9 genetic polymorphisms on the pharmacokinetics of losartan and its active metabolite, E-3174, through a systematic review and meta-analysis. Eight studies published before March 2021 were included in this study. We used
P C Wong et al.
The Journal of pharmacology and experimental therapeutics, 255(2), 584-592 (1990-11-01)
DuP 753 and PD123177 are two nonpeptide angiotensin II (AII)-specific ligands, which show high affinities for two respective and distinct subtypes of AII binding sites, i.e., AII-1 and AII-2 sites, respectively, in the rat adrenal gland, brain and uterus. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service