Skip to Content
Merck
All Photos(1)

Key Documents

SML2236

Sigma-Aldrich

L-012 sodium salt

≥98% (HPLC), powder, neurotoxin

Synonym(s):

8-Amino-5-chloro-2,3-dihydro-7-phenyl-Pyrido[3,4-d]pyridazine-1,4-dione, sodium salt, 8-Amino-5-chloro-7-phenyl-2,3-dihydro-pyrido[3,4-d]pyridazine-1,4-dione, sodium salt, L012

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8ClN4O2 · Na
CAS Number:
Molecular Weight:
310.67
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

L-012 sodium salt, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[Na+].Clc1nc(c(c3c1c(n[nH][c]3=O)[O-])N)c2ccccc2

InChI

1S/C13H9ClN4O2.Na/c14-11-8-7(12(19)17-18-13(8)20)9(15)10(16-11)6-4-2-1-3-5-6;/h1-5H,15H2,(H,17,19)(H,18,20);/q;+1/p-1

InChI key

IGEUYSJHQQCEFP-UHFFFAOYSA-M

Application

L-012 has been used as a probe in a bioluminescent assay for the determination of the reactive oxygen species (ROS) levels in the isolated mouse lungs.

Biochem/physiol Actions

L-012 is a luminol analog and a widely used reactive oxygen and nitrogen species (RONS; ROS & RNS) chemiluminescence (CL) probe both in cultures and in animals in vivo. L-012 displays significantly higher CL yield and sensitivity than luminol, lucigenin and MCLA. Instead of reacting with superoxide anion (O2) directly, L-012 (LumH2) is converted via a one-electron oxidation (catalyzed by peroxidase in the presence of H2O2) to the LumH· radical form, which then reacts with oxygen (O2) to yield O2 and L-012 quinone (q-Lum). The O2 in turn reacts with the LumH· radical, leading eventually to an endoperoxide that decomposes to emit luminescence. To a less extend, H2O2 can also react with q-Lum to yield luminescence. Although not specific to NADPH oxidase-derived RONS, L-012 is also commonly used for screening NADPH oxidase inhibitors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Nishinaka et al.
Biochemical and biophysical research communications, 193(2), 554-559 (1993-06-15)
A sensitive chemiluminescence method for measuring the production of superoxide anion (O2-) by activated EoL-1 cells (human eosinophilic leukemia cell line) is described. Recently, we succeeded in synthesizing a new chemiluminescence probe, 8-amino-5-chloro-7-phenylpyrido[3,4-d]pyridazine-1,4(2H,3H)dione (L-012). In the presence of L-012, activated
Muthusaravanan Sivaramakrishnan et al.
Current protocols, 3(8), e869-e869 (2023-08-25)
The reactive oxygen species (ROS) burst assay is a valuable tool for studying pattern-triggered immunity (PTI) in plants. During PTI, the interaction between pathogen recognition receptors (PRRs) and pathogen-associated molecular patterns (PAMPs) leads to the rapid production of ROS in
Peishan Li et al.
Nature communications, 11(1), 4387-4387 (2020-09-03)
The role of neutrophils in solid tumor metastasis remains largely controversial. In preclinical models of solid tumors, both pro-metastatic and anti-metastatic effects of neutrophils have been reported. In this study, using mouse models of breast cancer, we demonstrate that the
M Ii et al.
Biochemical and biophysical research communications, 193(2), 540-545 (1993-06-15)
An enhanced chemiluminescence reaction has been incorporated into an enzyme immunoassay (EIA) for human basic fibroblast growth factor (hbFGF). We developed a new luminol derivative, designated L-012 and a new enhancer, 4-(4-hydroxyphenyl)thiazole. Using these compounds, the detection limit of hbFGF
Andreas Daiber et al.
Redox biology, 12, 35-49 (2017-02-18)
Reactive oxygen and nitrogen species (RONS such as H2O2, nitric oxide) confer redox regulation of essential cellular functions (e.g. differentiation, proliferation, migration, apoptosis), initiate and catalyze adaptive stress responses. In contrast, excessive formation of RONS caused by impaired break-down by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service