Skip to Content
Merck
All Photos(1)

Key Documents

SML2233

Sigma-Aldrich

XMU-MP-1 hydrochloride

≥98% (HPLC)

Synonym(s):

4-((5,10-Dimethyl-6-oxo-6,10-dihydro-5H-pyrimido[5,4-b]thieno[3,2-e][1,4]diazepin-2-yl)amino)benzenesulfonamide hydrochloride, 4-[(6,10-Dihydro-5,10-dimethyl-6-oxo-5H-pyrimido[5,4-b]thieno[3,2-e][1,4]diazepin-2-yl)amino]benzenesulfonamide hydrochoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N6O3S2 · xHCl
CAS Number:
Molecular Weight:
416.48 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

CN(C1=CN=C(NC2=CC=C(S(N)(=O)=O)C=C2)N=C1N(C)C3=C4SC=C3)C4=O

InChI

1S/C17H16N6O3S2/c1-22-12-7-8-27-14(12)16(24)23(2)13-9-19-17(21-15(13)22)20-10-3-5-11(6-4-10)28(18,25)26/h3-9H,1-2H3,(H2,18,25,26)(H,19,20,21)

InChI key

YRDHKIFCGOZTGD-UHFFFAOYSA-N

Biochem/physiol Actions

Potent, reversible, ATP-competitive pan-MST kinase inhibitor with in vivo liver and intestinal tissue repair/regeneration efficacy.
XMU-MP-1 inhibits MST kinase activity (IC50 = 9.8 nM/MST1, 18.2 nM/MST2, 44.8 nM/MST3, 27.3 nM/MST4) in a reversible and ATP-competitive manner (MST1 IC50/[ATP] = 164 nM/10 μM and 4036 nM/300 μM; MST2 IC50/[ATP] = 34 nM/10 μM and 1498 nM/300 μM), exhibiting significant affinity and/or inhibitory potency toward only 17 other kinases among a panel of 468 (401 unique kinases). XMU-MP-1 selectively inhibits H2O2-stimulated MST autophosphorylation and phosphorylation of endogenous MST1/2 substrates (MOB1, LATS, YAP), but not JNK, in human and murine cells (Effective conc. 1 μM), effectively upregulating YAP nuclear localization and protecting HepG2 cells (3 μM) against MST2 overexpression-induced cell death. XMU-MP-1 exhibits in vivo efficacy toward liver and intestinal repair and regeneration in various murine models (1-3 mg/kg/day i.p.) and oral availability in rats (t1/2 = 5.18 h, Tmax = 3 h, AUC = 993 ng•h/mL, F = 39.48%; 10 mg/mL p.o.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Masashi Kanayama et al.
Nature immunology, 18(9), 973-984 (2017-07-04)
The balance of myeloid populations and lymphoid populations must be well controlled. Here we found that osteopontin (OPN) skewed this balance during pathogenic conditions such as infection and autoimmunity. Notably, two isoforms of OPN exerted distinct effects in shifting this
Hiroshi Kishimoto et al.
iScience, 26(7), 107146-107146 (2023-07-17)
Non-canonical Wnt signaling activated by Wnt5a/Wnt11 is required for the second heart field development in mice. However, the pathophysiological role of non-canonical Wnt signaling in the adult heart has not been fully elucidated. Here we show that cardiomyocyte-specific Wnt5a knockout
Fuqin Fan et al.
Science translational medicine, 8(352), 352ra108-352ra108 (2016-08-19)
Tissue repair and regenerative medicine address the important medical needs to replace damaged tissue with functional tissue. Most regenerative medicine strategies have focused on delivering biomaterials and cells, yet there is the untapped potential for drug-induced regeneration with good specificity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service