Skip to Content
Merck
All Photos(3)

Documents

SML0534

Sigma-Aldrich

Verteporfin

≥94% (HPLC), powder, YAP-TEA domain interaction inhibitor

Synonym(s):

(4R,4aS)-rel-18-ethenyl-4,4a-dihydro-3,4-bis(methoxycarbonyl)-4a,8,14,19-tetramethyl-24H,26H-Benzo[b]porphine-9,13-dipropanoic acid monomethyl ester, Visudyne, trans-18-ethenyl-4,4a-dihydro-3,4-bis(methoxycarbonyl)-4a,8,14,19-tetramethyl-23H,25H-Benzo[b]porphine-9,13-dipropanoic acid monomethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H42N4O8
CAS Number:
Molecular Weight:
718.79
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Verteporfin, ≥94% (HPLC)

Assay

≥94% (HPLC)

form

powder

storage condition

desiccated
protect from light

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

CC(C(/C=C1[C@@]2(C)C(/C(N/1)=C/3)=CC=C(C(OC)=O)[C@H]2C(OC)=O)=N/4)=C(CCC(OC)=O)C4=C\C5=C(CCC(O)=O)C(C)=C(/C=C6C(C=C)=C(C)C3=N/6)N5.CC(C(/C=C7[C@@]8(C)C(/C(N/7)=C/9)=CC=C(C(OC)=O)[C@H]8C(OC)=O)=N/%10)=C(CCC(O)=O)C%10=C\C%11=C(CCC(OC)=O)C(C)=C(/C=C%12C(C=C)

InChI

1S/2C41H42N4O8/c1-9-23-20(2)29-17-34-27-13-10-26(39(49)52-7)38(40(50)53-8)41(27,5)35(45-34)19-30-22(4)24(11-14-36(46)47)32(44-30)18-33-25(12-15-37(48)51-6)21(3)28(43-33)16-31(23)42-29;1-9-23-20(2)29-17-34-27-13-10-26(39(49)52-7)38(40(50)53-8)41(27,5)35(45-34)19-30-22(4)25(12-15-37(48)51-6)33(44-30)18-32-24(11-14-36(46)47)21(3)28(43-32)16-31(23)42-29/h2*9-10,13,16-19,38,43,45H,1,11-12,14-15H2,2-8H3,(H,46,47)/b31-16-,32-18-,34-17-,35-19-;31-16-,33-18-,34-17-,35-19-/t2*38-,41+/m00/s1

InChI key

YHNBVDZVUQFVLS-SKJZPIBWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Verteporfin has been used as a photosensitizer. It is also used as an inhibitor of YAP (Yes-associated protein)-TEA domain (TEAD) interaction.

Biochem/physiol Actions

Verteporfin has an ability to disrupt the interaction between YAP (Yes-associated protein)/TAZ (transcriptional coactivator with PDZ-binding motif) and TEA domain (TEAD) complex. It also reduces the viability of the ovarian cancer cells and almost eliminates cell migration. Therefore, verteporfin might be considered as a potent therapeutic for ovarian cancer.
Verteporfin is a photosensitizer for photodynamic therapy to eliminate the abnormal blood vessels in the eye associated with conditions such as macular degeneration. Verteporfin accumulates in abnormal blood vessels and, when stimulated by nonthermal red light with a wavelength of 693 nm in the presence of oxygen, produces highly reactive short-lived singlet oxygen and other reactive oxygen radicals, resulting in local damage to the endothelium and blockage of the vessels. Verteporfin localizes predominantly in mitochondria.

Other Notes

Light sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akihiro Yamashita et al.
Stem cells translational medicine, 10(1), 115-127 (2020-08-22)
Human induced pluripotent stem cells (hiPSCs) are a promising cell source for the creation of cartilage to treat articular cartilage damage. The molecular mechanisms that translate culture conditions to the chondrogenic differentiation of hiPSCs remain to be analyzed. To analyze
YAP1 and AR interactions contribute to the switch from androgen-dependent to castration-resistant growth in prostate cancer.
Kuser-Abali G, et al.
Nature Communications, 6, 8126-8126 (2015)
Yukari Mae et al.
Molecular and clinical oncology, 13(3), 10-10 (2020-08-06)
Photodynamic therapy (PDT) induces photochemical reactions, resulting in the destruction of tumor cells via singlet (S1) oxygen production. This cellular destruction occurs specifically in tumor cells, following selective accumulation of a photosensitizer and its excitation by a specific wavelength. Verteporfin
Alan F Cruess et al.
Acta ophthalmologica, 87(2), 118-132 (2008-06-26)
Photodynamic therapy (PDT) with verteporfin has been used less comprehensively in the treatment of exudative age-related macular degeneration (AMD), and specifically of choroidal neovascularization (CNV), since the advent of antiangiogenic therapies. Recently, there has been a renewed interest in PDT
Victor C K Lo et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 31(9), 1398-1405 (2013-04-30)
Photodynamic therapy (PDT) has been shown to ablate tumors within vertebral bone and yield short-term improvements in vertebral architecture and biomechanical strength, in particular when combined with bisphosphonate (BP) treatment. Longer-term outcomes of PDT combined with current treatments for skeletal

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service