Skip to Content
Merck
All Photos(1)

Key Documents

529573

Sigma-Aldrich

PP2

≥95% (HPLC), solid, Src protein tyrosine kinase inhibitor, Calbiochem®

Synonym(s):

PP2, AG 1879, 4-Amino-5-(4-chlorophenyl)-7-( t-butyl)pyrazolo[3,4-d]pyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16ClN5
CAS Number:
Molecular Weight:
301.77
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

PP2, PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC₅₀ = 4, 5, 5, &100 nM for p56lck, p59fynT, Hck, & Src, respectively).

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 20 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Clc1ccc(cc1)c2n[n](c3ncnc(c32)N)C(C)(C)C

InChI

1S/C15H16ClN5/c1-15(2,3)21-14-11(13(17)18-8-19-14)12(20-21)9-4-6-10(16)7-5-9/h4-8H,1-3H3,(H2,17,18,19)

InChI key

PBBRWFOVCUAONR-UHFFFAOYSA-N

General description

A potent, reversible, ATP-competitive, and selective inhibitor of the Src family of protein tyrosine kinases. Inhibits p56lck (IC50 = 4 nM), p59fynT (IC50 = 5 nM), Hck (IC50 = 5 nM), and Src (IC50 = 100 nM). Does not significantly affect the activity of EGFR kinase (IC50 = 480 nM), JAK2 (IC50 >50 µM), or ZAP-70 (IC50 >100 µM). Inhibits the activation of focal adhesion kinase and its phosphorylation at Tyr577. Also potently inhibits anti-CD3-stimulated tyrosine phosphorylation of human T cells (IC50 = 600 nM). A 10 mM (1 mg/331 µl) solution of PP2 (Cat. No. 529576) in DMSO also available.

Biochem/physiol Actions

Cell permeable: no
Primary Target
p56lck
Product competes with ATP.
Reversible: yes
Target IC50: 4 nM, 5 nM, 5 nM, 100 nM, against p56lck, p59fynT, Hck, and Src, respectively; 600 nM against anti-CD3-stimulated tyrosine phosphorylation of human T cells

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

Further dilute with aqueous buffers just prior to use.

Other Notes

Karni, R., et al. 2003. FEBS Lett.537, 47.
Salazar, E.P., and Rozengurt, E. 1999. J. Biol. Chem. 274, 28371.
Hanke, J.H., et al. 1996. J. Biol. Chem.271, 695.



Selected Citations
Lee, J., et al. 2009. Cell Stem Cell5, 76.


Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Yuko Murakami-Nishimagi et al.
Breast cancer research : BCR, 25(1), 41-41 (2023-04-15)
Cell adhesion is indispensable for appropriate tissue architecture and function in multicellular organisms. Besides maintaining tissue integrity, cell adhesion molecules, including tight-junction proteins claudins (CLDNs), exhibit the signaling abilities to control a variety of physiological and pathological processes. However, it
Elliot Dine et al.
Cell reports, 35(12), 109280-109280 (2021-06-24)
Protein clustering is pervasive in cell signaling, yet how signaling from higher-order assemblies differs from simpler forms of molecular organization is still poorly understood. We present an optogenetic approach to switch between oligomers and heterodimers with a single point mutation.
Emily C Suter et al.
Cell reports, 36(8), 109587-109587 (2021-08-26)
Cancer immunotherapies often modulate macrophage effector function by introducing either targeting antibodies that activate Fcγ receptors (FcγRs) or blocking antibodies that disrupt inhibitory SIRPα-CD47 engagement. However, how these competing signals are integrated is poorly understood, raising questions about how to
Zahra Alvandi et al.
Methods in molecular biology (Clifton, N.J.), 2520, 261-264 (2021-10-07)
Embryonic stem cells (ESCs) are a unique model that allows the study of molecular pathways underlying commitment and differentiation. We have studied signaling pathways and their contributions to osteogenic differentiation. In addition to our previously published protocol where we recommended
Anna M Lone et al.
Science signaling, 14(703), eabc8579-eabc8579 (2021-10-06)
Prostaglandin E2 (PGE2) promotes an immunosuppressive microenvironment in cancer, partly by signaling through four receptors (EP1, EP2, EP3, and EP4) on T cells. Here, we comprehensively characterized PGE2 signaling networks in helper, cytotoxic, and regulatory T cells using a phosphoproteomics

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service