Skip to Content
Merck
All Photos(1)

Documents

SML1747

Sigma-Aldrich

JNJ-55511118

≥98% (HPLC)

Synonym(s):

5-[2-Chloro-6-(trifluoromethoxy)phenyl]-1,3-dihydrobenzimidazol-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8ClF3N2O2
Molecular Weight:
328.67
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

JNJ-55511118 is also known as 5-[2-chloro-6-(trifluoromethoxy)phenyl]-1,3-dihydrobenzimidazol-2-one. It acts as an anticonvulsant or neuroprotectant. It is considered as a reversible α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA) receptor inhibitor, mainly in the hippocampus region.
JNJ-55511118 is an orally available, brain penetrant, potent and selective negative modulator of AMPA receptors that requires the presence of the accessory protein CACNG8 (TARP-γ8). JNJ-55511118 exhibit potent inhibition of neurotransmission within hippocampus, and a strong anti-convulsant effect.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael P Maher et al.
The Journal of pharmacology and experimental therapeutics, 357(2), 394-414 (2016-03-19)
Members of the α-amino-3-hydroxyl-5-methyl-4-isoxazole-propionic acid (AMPA) subtype of ionotropic glutamate receptors mediate the majority of fast synaptic transmission within the mammalian brain and spinal cord, representing attractive targets for therapeutic intervention. Here, we describe novel AMPA receptor modulators that require

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service