PHL10048
4-Monobromocannabidiol
phyproof® Reference Substance
Synonym(s):
4-MBO-CBD
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥90.0% (HPLC)
manufacturer/tradename
PhytoLab
application(s)
food and beverages
pharmaceutical
storage temp.
2-8°C
General description
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
This substance is tested for its content of d8-THC, d9-THC, and d9-THCA. Results are given on each COA. Acceptable maximum limits were set to 0.10 % each, and to 0.20 % for the sum of d9-THC and d9-THCA (in analogy to the cannabidiol monograph in DAC).
This substance is tested for its content of d8-THC, d9-THC, and d9-THCA. Results are given on each COA. Acceptable maximum limits were set to 0.10 % each, and to 0.20 % for the sum of d9-THC and d9-THCA (in analogy to the cannabidiol monograph in DAC).
Legal Information
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Repr. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service