Skip to Content
Merck
All Photos(2)

Key Documents

P5295

Sigma-Aldrich

Piracetam

Synonym(s):

2-(2-Oxopyrrolidino)acetamide, 2-Oxo-1-pyrrolidineacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N2O2
CAS Number:
Molecular Weight:
142.16
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

form

powder

SMILES string

NC(=O)CN1CCCC1=O

InChI

1S/C6H10N2O2/c7-5(9)4-8-3-1-2-6(8)10/h1-4H2,(H2,7,9)

InChI key

GMZVRMREEHBGGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Piracetam has been used to study crystal structure and physicochemical properties of six co-crystals of piracetam. It has also been used to study its in vitro antioxidant properties.

Biochem/physiol Actions

Piracetam (2-oxo-1-pyrrolidinyl-acetamide) is a nootropic drug, which enhances memory and facilitates learning. Piracetam also acts as a vasodilator and improves blood flow. It has a potential to treat cognitive impairment in aging, brain injury, memory and balance problems. In addition, piracetam is also used to treat peripheral vascular disease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Piracetam and vinpocetine ameliorate rotenone-induced Parkinsonism in rats
Zaitone SA, et al.
International Journal of Pharmaceutics, 44(6), 774-774 (2012)
A H Gouliaev et al.
Brain research. Brain research reviews, 19(2), 180-222 (1994-05-01)
Nearly three decades have now passed since the discovery of the piracetam-like nootropics, compounds which exhibit cognition-enhancing properties, but for which no commonly accepted mechanism of action has been established. This review covers clinical, pharmacokinetic, biochemical and behavioural results presented
In vitro antioxidant properties of pentoxifylline, piracetam, and vinpocetine
Horvath B, et al.
Clinical neuropharmacology, 25(1), 37-42 (2002)
Piracetam co-crystals with OH-group functionalized carboxylic acids
Viertelhaus M, et al.
Crystal Growth & Design, 9(5), 2220-2228 (2009)
Measurement and correlation for the solubilities of cinnarizine, pentoxifylline, and piracetam in supercritical carbon dioxide
Lee, C, et al.
Fluid Phase Equilibria, 367(1), 182-187 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service