Skip to Content
Merck
All Photos(1)

Key Documents

SML1135

Sigma-Aldrich

MG-132(R)

≥95% (HPLC)

Synonym(s):

Z-L-Leu-D-Leu-L-Leu-al

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H41N3O5
CAS Number:
Molecular Weight:
475.62
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

product line

SAFC Hitech®

Quality Level

Assay

≥95% (HPLC)

form

powder

solubility

DMSO: soluble

storage temp.

−20°C

SMILES string

O=C(N[C@H](CC(C)C)C(N[C@H](C=O)CC(C)C)=O)[C@H](CC(C)C)NC(OCC1=CC=CC=C1)=O

InChI

1S/C26H41N3O5/c1-17(2)12-21(15-30)27-24(31)22(13-18(3)4)28-25(32)23(14-19(5)6)29-26(33)34-16-20-10-8-7-9-11-20/h7-11,15,17-19,21-23H,12-14,16H2,1-6H3,(H,27,31)(H,28,32)(H,29,33)/t21-,22+,23-/m0/s1

InChI key

TZYWCYJVHRLUCT-ZRBLBEILSA-N

Gene Information

Biochem/physiol Actions

MG-132(R) is a potent, membrane-permeable proteasome inhibitor. It induces neurite outgrowth in PC12 cells at 10 M. MG-132(R) blocks cleavage of poly(ADP-ribose) polymerase and apoptosis in thymocytes. However, MG-132(R) also activates c-Jun N-terminal protein kinase (JNK-1), which initiates apoptosis in response to cell stress. Proteasome inhibition induces accumulation of heat shock protein mRNA, activation of heat-shock proteins, and enhanced thermotolerance in various cell types.

Legal Information

SAFC Hitech is a registered trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xueyan Zhao et al.
Developmental and comparative immunology, 106, 103632-103632 (2020-01-29)
Tightly regulation of NF-κB signaling is essential to innate and adaptive immune responses, but its regulatory mechanism remains unclear in various organisms, especially teleost fish. In this study, we reported that IRF3 attenuates the inhibitory effect of IκBα on NF-κB
Fanjie Jin et al.
Oncology letters, 19(1), 858-868 (2020-01-04)
The clinical significance of the proteasome inhibitor MG132 has been examined in numerous human cancer types; however, its influence on the metastasis and progression of pancreatic cancer is yet to be determined. In the present study, the effect of MG132
The effect of MG132, a proteasome inhibitor on HeLa cells in relation to cell growth, reactive oxygen species and GSH.
Han YH
Oncology Reports, 22(1), 215-221 (2009)
Ling-Yun Chu et al.
Scientific reports, 7(1), 12472-12472 (2017-10-01)
Pro-inflammatory cytokines are known to induce endothelial cell autophagy, but the role of autophagy in regulating the expression of pro-inflammatory molecules has not been characterized. We hypothesized that autophagy facilitates expression of endothelial adhesion molecules. TNFα and IL-1β induced autophagy
Shashipavan Chillappagari et al.
Journal of cystic fibrosis : official journal of the European Cystic Fibrosis Society, 20(1), 140-148 (2020-06-15)
The stress-regulated enzyme hemeoxygenase-1 (HO-1) contributes to the cell response towards inflammation and oxidative stress. We previously reported on curtailed HO-1 expression in cystic fibrosis (CF) bronchial epithelial (CFBE41o-) cells and CF-mice, but the molecular mechanisms for this are not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service