Skip to Content
Merck
All Photos(1)

Key Documents

C4859

Sigma-Aldrich

Cycloheximide solution

Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture

Synonym(s):

CHX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NO4
CAS Number:
Molecular Weight:
281.35
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.77

biological source

microbial

Quality Level

description

suitable for testing specifications (CHO C. growth inhibitor)

form

solution

concentration

100 mg/mL in DMSO

solubility

DMSO: soluble

antibiotic activity spectrum

fungi
yeast

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

O=C1[C@@]([C@H](O)CC2CC(NC(C2)=O)=O)([H])C[C@@H](C)C[C@@H]1C

InChI

1S/C15H23NO4/c1-8-3-9(2)15(20)11(4-8)12(17)5-10-6-13(18)16-14(19)7-10/h8-12,17H,3-7H2,1-2H3,(H,16,18,19)/t8-,9-,11-,12+/m0/s1

InChI key

YPHMISFOHDHNIV-FSZOTQKASA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Specificity

Tested for cell growth arrest, selection of cycloheximide resistant yeast, apoptosis induction, and facilitation of apoptosis induction by FasL.

Biochem/physiol Actions

Cycloheximide (CHX) is an antibiotic produced by S. griseus. Its main biological activity is translation inhibition in eukaryotes resulting in cell growth arrest and cell death. CHX is widely used for selection of CHX-resistant strains of yeast and fungi, controlled inhibition of protein synthesis for detection of short-lived proteins and super-induction of protein expression, and apoptosis induction or facilitation of apoptosis induction by death receptors.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.Hygroscopic. Storage class (TRGS 510): Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye-Yeoung Yun et al.
PLoS biology, 17(7), e3000367-e3000367 (2019-07-20)
Human papillomaviruses (HPVs) are causative agents of various diseases associated with cellular hyperproliferation, including cervical cancer, one of the most prevalent tumors in women. E7 is one of the two HPV-encoded oncoproteins and directs recruitment and subsequent degradation of tumor-suppressive
Konstantinos Boulias et al.
Molecular cell, 75(3), 631-643 (2019-07-08)
mRNAs are regulated by nucleotide modifications that influence their cellular fate. Two of the most abundant modified nucleotides are N6-methyladenosine (m6A), found within mRNAs, and N6,2'-O-dimethyladenosine (m6Am), which is found at the first transcribed nucleotide. Distinguishing these modifications in mapping
Anna Duarri et al.
Annals of neurology, 72(6), 870-880 (2013-01-03)
To identify the causative gene for the neurodegenerative disorder spinocerebellar ataxia type 19 (SCA19) located on chromosomal region 1p21-q21. Exome sequencing was used to identify the causal mutation in a large SCA19 family. We then screened 230 ataxia families for
Xing Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(43), 15438-15443 (2014-10-16)
The receptor-interacting kinase-3 (RIP3) and its downstream substrate mixed lineage kinase domain-like protein (MLKL) have emerged as the key cellular components in programmed necrotic cell death. Receptors for the cytokines of tumor necrosis factor (TNF) family and Toll-like receptors (TLR)
V O Kaminskyy et al.
Cell death & disease, 4, e522-e522 (2013-03-09)
TNF-related apoptosis-inducing ligand (TRAIL) is a promising cytokine for killing tumor cells. However, a number of studies have demonstrated that different cancer cells resist TRAIL treatment and, moreover, TRAIL can promote invasion and metastasis in resistant cells. Here we report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service