Skip to Content
Merck
All Photos(1)

Key Documents

5.04314

Sigma-Aldrich

Bortezomib

≥98% (LC/MS), solid, 20S proteasome inhibitor, Calbiochem®

Synonym(s):

Bortezomib, (R)-3-Methyl-1-((S)-3-phenyl-2-(pyrazine-2-carboxamido)propanamido)butylboronic acid, BTZ, LDP-341, LDP341, MG341, MLN-341, MLN341, PS341, Proteasome Inhibitor XXII, PS-341, MG-341, Pyz-Phe-boroLeu, BTZ, LDP-341, LDP341, MG341, MLN-341, MLN341, PS341, Proteasome Inhibitor XXII, PS-341, MG-341, Pyz-Phe-boroLeu, (R)-3-Methyl-1-((S)-3-phenyl-2-(pyrazine-2-carboxamido)propanamido)butylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25BN4O4
CAS Number:
Molecular Weight:
384.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Bortezomib,

Assay

≥98% (LC/MS)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white

solubility

DMSO: 100 mg/mL
ethanol: 2 mg/mL (with sonication)

storage temp.

−20°C

SMILES string

B(O)(O)[C@@H](NC(=O)[C@@H](NC(=O)c2nccnc2)Cc1ccccc1)CC(C)C

InChI

1S/C19H25BN4O4/c1-13(2)10-17(20(27)28)24-18(25)15(11-14-6-4-3-5-7-14)23-19(26)16-12-21-8-9-22-16/h3-9,12-13,15,17,27-28H,10-11H2,1-2H3,(H,23,26)(H,24,25)/t15-,17-/m0/s1

InChI key

GXJABQQUPOEUTA-RDJZCZTQSA-N

General description

Bortezomib, a cell-permeable dipeptidylboronate compound, is a proteasome inhibitor. The proteasomal system is crucial for cellular protein turnover, which is necessary for maintaining cell homeostasis. Bortezomib binds reversibly to the chymotrypsin-like subunit of the 26S proteasome, inhibiting its function and thereby preventing the degradation of multiple pro-apoptotic factors. Bortezomib selectively inhibits 20S proteasome β5 ChTL/chymotrypsin- over β1 CL/caspase- and β2 TL/trypsin-like activity (kinact/Ki = 38,000, 5,700, and <100 M-1s-1, respectively, in human 20S proteasome assays using 10 M Suc-LLVY-AMC/Cat. No. 539142, 10 M Z-LLE-AMC/Cat. No. 539141, or 50 M Boc-LRR-AMC as substrate; IC50 in 1 h = 7, 74, and 4,200 nM, respectively) via a covalent, slowly reversible, interaction between the nucleophilic Thr1 hydroxy group/Thr1Oγ of the catalytic β subunit and the inhibitor′s electrophilic boronic moiety, displaying much reduced potency against human chymotrypsin, cathepsin G, leukocyte elastase, and thrombin (Ki = 0.32, 0.63, 2.3, and 13 M, respectively, vs. 620 nM using rabbit muscle 20S). A widely used inhibitor both in cultures in vitro and in animals in vivo. Despite being the first proteasome inhibitor approved by FDA for clinical anticancer treatment, its therapeutic efficacy continues to be hampered by off-target effects and dose-limiting toxicity.

Application

Bortezomib has been used to induce aerobic glycolysis/ neuropathic pain in mice and study chemotherapy-induced painful peripheral neuropathy.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Features and Benefits

  • Cell permeable and enables targeted action
  • Allows reversible modulation of cellular processes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

Use only fresh DMSO or Ethanol for reconstitution.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C.) Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Du, X.L, and Chen, Q. 2013. Acta Haematol.129, 207.
Tamatani, T., et al. 2013. Int. J. Oncol.42, 935.
Beck, P., et al. 2012. J. Biol. Chem.393, 1101.
Fang, H.T., et al. 2012. Proc. Natl. Acad. Sci. USA.109, 2521.
Chen, D., et al. 2011. Curr. Cancer Drug Targets11, 239.
Demo, S.D., et al. 2007. Cancer Res.67, 6383.
Adams, J., et al. 1999. Cancer Res.59, 2615.
Teicher, B.A., et al. 1999. Clin. Cancer Res.5, 2638.
Adams, J., et al. 1998. Bioorg. Med. Chem. Lett.8, 333.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susan D Demo et al.
Cancer research, 67(13), 6383-6391 (2007-07-10)
Clinical studies with bortezomib have validated the proteasome as a therapeutic target for the treatment of multiple myeloma and non-Hodgkin's lymphoma. However, significant toxicities have restricted the intensity of bortezomib dosing. Here we describe the antitumor activity of PR-171, a
Panjamurthy Kuppusamy et al.
Frontiers in pain research (Lausanne, Switzerland), 5, 1424348-1424348 (2024-07-09)
Chemotherapy-induced peripheral neuropathy (CIPN) is a debilitating side effect of cancer treatment that significantly impacts patients' quality of life. This study investigated the effects of targeting metabolic pathways on bortezomib-induced neuropathic pain and tumor growth using a Lewis lung carcinoma
B A Teicher et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 5(9), 2638-2645 (1999-09-28)
The anticancer activity of the boronic acid dipeptide proteasome inhibitor PS-341 was examined in vitro and in vivo. PS-341 was a potent cytotoxic agent toward MCF-7 human breast carcinoma cells in culture, producing an IC90 of 0.05 microM on 24
J Adams et al.
Bioorganic & medicinal chemistry letters, 8(4), 333-338 (1999-01-01)
Potent and selective dipeptidyl boronic acid proteasome inhibitors are described. As compared to peptidyl aldehyde compounds, boronic acids in this series display dramatically enhanced potency. Compounds such as 15 are promising new therapeutics for treatment of cancer and inflammatory diseases.
Hai-Tong Fang et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(7), 2521-2526 (2012-02-07)
The boronic acid dipeptide bortezomib inhibits the chymotrypsin-like activity of the 26S proteasome and shows significant therapeutic efficacy in multiple myeloma. However, recent studies suggest that bortezomib may have more complex mechanisms of action in treating cancer. We report here

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service