Skip to Content
Merck
All Photos(1)

Key Documents

SML0830

Sigma-Aldrich

SNAP-94847

≥98% (HPLC)

Synonym(s):

N-[3-[1-[[4-(3,4-Difluorophenoxy)phenyl]methyl]-4-piperidinyl]-4-methylphenyl]-2-methyl-propanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H32F2N2O2
CAS Number:
Molecular Weight:
478.57
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1c(ccc(c1)Oc2ccc(cc2)CN3CCC(CC3)c4c(ccc(c4)NC(=O)C(C)C)C)F.Cl

InChI

1S/C29H32F2N2O2.ClH/c1-19(2)29(34)32-23-7-4-20(3)26(16-23)22-12-14-33(15-13-22)18-21-5-8-24(9-6-21)35-25-10-11-27(30)28(31)17-25;/h4-11,16-17,19,22H,12-15,18H2,1-3H3,(H,32,34);1H

InChI key

DEDUDFNRQKUBRH-UHFFFAOYSA-N

Biochem/physiol Actions

SNAP-94847 is a potent antagonist of the melanin-concentrating hormone receptor MCHR-1.
SNAP-94847 is a potent antagonist of the melanin-concentrating hormone receptor MCHR-1. SNAP-94847 displays anxiolytic and antidepressant activity, and reduces food intake in mice.

Features and Benefits

This compound is featured on the Melanin-Concentrating Hormone Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service