Skip to Content
Merck
All Photos(1)

Key Documents

SMB00231

Sigma-Aldrich

Loganic acid

≥95% (LC/MS-ELSD)

Synonym(s):

Loganate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24O10
CAS Number:
Molecular Weight:
376.36
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

C[C@H]1[C@@H](O)C[C@H]2[C@@H]1[C@@H](OC=C2C(O)=O)O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O

InChI

1S/C16H24O10/c1-5-8(18)2-6-7(14(22)23)4-24-15(10(5)6)26-16-13(21)12(20)11(19)9(3-17)25-16/h4-6,8-13,15-21H,2-3H2,1H3,(H,22,23)/t5-,6+,8-,9+,10+,11+,12-,13+,15-,16-/m0/s1

InChI key

JNNGEAWILNVFFD-CDJYTOATSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Loganic acid is a naturally occurring iridoid monoterpeneglusoside. It is a key constituent of Swertia caroliniensis.
Natural product derived from plant source.

Biochem/physiol Actions

Loganic acid is involved in the biosynthesis of indole alkaloids in Vinca rosea.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Nannan Wang et al.
Biomedical chromatography : BMC, 32(6), e4195-e4195 (2018-01-20)
To reveal the material basis of Huo Luo Xiao Ling Dan (HLXLD), a sensitive and selective ultra-high performance liquid chromatography coupled with quadrupole-time-of-flight mass spectrometry (UHPLC-Q-TOF/MS) method was developed to identify the absorbed components and metabolites in rat plasma after
Rilka M Taskova et al.
Journal of natural products, 74(6), 1477-1483 (2011-05-17)
From an extract of Veronica (sect. Hebe) lavaudiana we have identified mannitol and isolated 11 iridoid glucosides, the carbohydrate ester hebitol II, and four phenylethanoid glycoside esters. Five of the iridoid glycosides are new; of these, lavaudiosides A, B, and
S-Adenosyl-l-methionine: Loganic Acid Methyltransferase A CARBOXYL-ALKYLATING ENZYME FROM Vinca rosea.
Madyastha K M, et al.
The Journal of Biological Chemistry, 248(7), 2497-2501 (1973)
Jie Li et al.
Frontiers in chemistry, 5, 125-125 (2018-01-10)
Roots, stems, leaves, and flowers of Longdan (
Lili Xu et al.
Journal of ethnopharmacology, 209, 91-99 (2017-07-25)
Swertia cincta Burkill was traditionally used for treating jaundice and various types of chronic and acute hepatitis in Yunnan and Tibet in China for hundreds of years. This study aims to investigate the protective effect of S. cincta Burkill (ESC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service