Skip to Content
Merck
All Photos(1)

Documents

SML0153

Sigma-Aldrich

Geniposide

≥98% (HPLC)

Synonym(s):

(1S,4αS,7αS)- 1-(β-D-Glucopyranosyloxy)-1,4a,5,7a-tetrahydro-7-(hydroxymethyl)- cyclopenta[c]pyran-4-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24O10
CAS Number:
Molecular Weight:
388.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: ≥5 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H]3[C@@H]1CC=C3CO

InChI

1S/C17H24O10/c1-24-15(23)9-6-25-16(11-7(4-18)2-3-8(9)11)27-17-14(22)13(21)12(20)10(5-19)26-17/h2,6,8,10-14,16-22H,3-5H2,1H3/t8-,10-,11-,12-,13+,14-,16+,17+/m1/s1

InChI key

IBFYXTRXDNAPMM-BVTMAQQCSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Geniposide is an iridoid glycoside with a variety of biological activities including neuroprotective, anti-diabetic, antiproliferative, and antioxidative activity. Geniposide has been shown to regulate Nrf2 translocation and upregulate downstream antioxidant protein HO-1 expression through the PI3K/Akt signaling pathway, which may account for its neuroprotective and antioxidative activity. Glucagon-like peptide 1 (GLP-1) receptor may also play a critical role in geniposide induced HO-1 expression.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian Li et al.
PloS one, 7(9), e45811-e45811 (2012-10-11)
Refined Qing-Kai-Ling (QKL), a modified Chinese medicine, consists of three main ingredients (Baicalin, Jasminoidin and Desoxycholic acid), plays a synergistic effect on the treatment of the acute stage of ischemic stroke. However, the rules of the combination and synergism are
Yan-Jing Li et al.
Rapid communications in mass spectrometry : RCM, 26(11), 1377-1384 (2012-05-05)
The Direct Analysis in Real Time (DART) ionization source coupled with a quadrupole time-of-flight tandem mass spectrometry (Q-TOF MS/MS) system has the capability to desorb analytes directly from samples from complex Chinese herbal preparations without sample cleanup or chromatographic separation.
Jun Wang et al.
Neurochemical research, 37(10), 2235-2248 (2012-08-08)
Geniposide, an iridoid glycoside isolated from Gardenia, has neuroprotective activities against oxidative stress and inflammation. The present study investigated the in vivo protective effect of geniposide on ischemia/reperfusion-injured rats by middle cerebral artery occlusion (MCAO), and the inhibitory effects of
K Hiramatsu et al.
Alimentary pharmacology & therapeutics, 36(6), 575-586 (2012-07-24)
Idiopathic mesenteric phlebosclerosis (IMP) is a rare disease, characterised by thickening of the wall of the right hemicolon with calcification of mesenteric veins. However, the aetiology remains unknown. To investigate the possible association of herbal medicines with IMP. The clinical
Jianhui Liu et al.
Toxicology in vitro : an international journal published in association with BIBRA, 26(7), 1093-1097 (2012-07-24)
β-Cell apoptosis is considered to be a major cause of loss of β cells in diabetes. Geniposide could prevent oxidative stress-induced neuron apoptosis, and improved glucose stimulated insulin secretion by activating glucagon-like peptide 1 receptor (GLP-1R) in INS-1 cells. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service