Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300100

Sigma-Aldrich

Anti-phospho-ATM (pSer1981) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AT1 antibody produced in rabbit, Anti-ATA antibody produced in rabbit, Anti-ATC antibody produced in rabbit, Anti-ATD antibody produced in rabbit, Anti-ataxia telangiectasia mutated antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~350 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100

isotype

IgG

immunogen sequence

(E-G-SP-Q-S)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer1981)

Gene Information

human ... ATM(472)

Immunogen

Peptide sequence around phosphorylation site of serine 1981 (E-G-S(p)-Q-S), according to the protein ATM.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

ATM encoded by this gene belongs to the PI3/PI4-kinase family. This protein is an important cell cyclecheckpoint kinase that phosphorylates; thus, it functions as a regulator of a wide variety of downstream proteins,including tumor suppressor proteins p53 and BRCA1, checkpoint kinase CHK2, checkpoint proteins RAD17 and RAD9, and DNArepair protein NBS1. This protein and the closely related kinase ATR are thought to be master controllers of cellcycle checkpoint signaling pathways that are required for cell response to DNA damage and for genome stability.Mutations in this gene are associated with ataxia telangiectasia, an autosomal recessive disorder. Two transcriptvariants encoding different isoforms have been found for this gene.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacinta H Martin et al.
Scientific reports, 8(1), 9685-9685 (2018-06-28)
The unique biology of the oocyte means that accepted paradigms for DNA repair and protection are not of direct relevance to the female gamete. Instead, preservation of the integrity of the maternal genome depends on endogenous protein stores and/or mRNA
Natalia Konieczna et al.
International journal of molecular sciences, 20(11) (2019-06-04)
Human telomeres were one of the first discovered and characterized sequences forming quadruplex structures. Association of these structures with oncogenic and tumor suppressor proteins suggests their important role in cancer development and therapy efficacy. Since cationic porphyrin TMPyP4 is known
Xiao Long et al.
Journal of cellular biochemistry, 120(4), 5652-5665 (2018-11-06)
Osteosarcoma (OS) is a primary malignant bone tumor with high morbidity. Developing new therapeutic approaches with neoadjuvant is of great interest in OS treatment. Reportedly, ataxia telangiectasia mutated (ATM)/ataxia telangiectasia and radiation resistance gene 3 related (ATR)-p53 signaling is considered
Nan Li et al.
OncoTargets and therapy, 12, 3065-3075 (2019-05-23)
Human males absent on the first (hMOF) is a histone acetyltransferase (HAT) and is responsible for acetylating histone H4 at lysine 16 (H4K16). Recent studies have indicated that hMOF is overexpressed in non-small-cell lung cancer (NSCLC) as an oncogene. The
Pinakin Pandya et al.
Cellular signalling, 62, 109340-109340 (2019-06-09)
Protein kinase C (PKC)-interacting cousin of thioredoxin (PICOT; also termed glutaredoxin 3 (Glrx3)) is a ubiquitously expressed protein that possesses an N-terminal monothiol thioredoxin (Trx) domain and two C-terminal tandem copies of a monothiol Glrx domain. It has an overall

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service