Skip to Content
Merck
All Photos(8)

Key Documents

SAB4504476

Sigma-Aldrich

Anti-phospho-mTOR (pSer2448) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 288 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer2448)

Gene Information

human ... MTOR(2475)

General description

The mechanistic/mammalian target of rapamycin (mTOR) is a serine/threonine kinase. It is a member of the phosphatidylinositide 3-kinases (PI 3-kinases or PI3Ks) family. It is mapped to human chromosome 1p36.22.

Immunogen

The antiserum was produced against synthesized peptide derived from human mTOR around the phosphorylation site of Ser2448.

Immunogen Range: 2415-2464

Application

Anti-phospho-mTOR (pSer2448) antibody has been used in western blotting.

Biochem/physiol Actions

The mechanistic/mammalian target of rapamycin (mTOR) controls protein translation, cell growth, survival, invasion and metabolism. It acts as an intracellular signalling regulator, which links the extracellular events to the intracellular process. mTOR also acts on the protein synthesis machinery.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Yuan et al.
International journal of molecular medicine, 41(4), 2086-2098 (2018-01-19)
Presbycusis is the most common sensory impairment associated with aging; however, the underlying molecular mechanism remains unclear. Autophagy has been demonstrated to serve a key role in diverse diseases; however, no studies have examined its function in central presbycusis. The
Cap-independent translation ensures mTOR expression and function upon protein synthesis inhibition
Marques-Ramos A, et al.
RNA, rna-063040 (2017)
Yulin Zhao et al.
Aging, 13(18), 22459-22473 (2021-09-24)
Emerging studies have revealed that non-coding RNAs contribute to regulating intervertebral disc degeneration (IVDD). Here, we intended to probe into the function of miR-19b-3p in IVDD evolvement. The miR-19b-3p level in the intervertebral disc (IVD) tissues of IVDD patients and
Autophagy regulates the degeneration of the auditory cortex through the AMPK-mTOR-ULK1 signaling pathway
Yuan J, et al.
International Journal of Molecular Medicine, 41(4), 2086-2098 (2018)
Impaired Cellular Energy Metabolism Contributes to Duck-Enteritis-Virus-Induced Autophagy via the AMPK-TSC2-MTOR Signaling Pathway
Yin H, et al.
Frontiers in Cellular and Infection Microbiology, 7(8), 423-423 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service