Skip to Content
Merck
All Photos(1)

Key Documents

SAB2102033

Sigma-Aldrich

Anti-RNF43 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp781H02126, Anti-DKFZp781H0392, Anti-FLJ20315, Anti-MGC125630, Anti-Ring finger protein 43

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

86 kDa

species reactivity

mouse, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF43(54894)

Immunogen

Synthetic peptide directed towards the middle region of human RNF43

Biochem/physiol Actions

RNF43 is a HAP95 (AKAP8L) binding ubiquitin ligase that promotes cell growth and is upregulated in colon cancer.

Sequence

Synthetic peptide located within the following region: DFDPLVYCSPKGDPQRVDMQPSVTSRPRSLDSVVPTGETQVSSHVHYHRH

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunhe Gao et al.
Stem cell research & therapy, 8(1), 98-98 (2017-04-28)
Ring finger protein 43 (RNF43) is a member of the transmembrane E3 ubiquitin ligase family that was originally found in stem cells and plays important roles in tumor formation and progression. Our previous study indicated that RNF43 might be a
H Nailwal et al.
Cell death & disease, 6, e1768-e1768 (2015-05-23)
The interplay between influenza virus and host factors to support the viral life cycle is well documented. Influenza A virus (IAV) proteins interact with an array of cellular proteins and hijack host pathways which are at the helm of cellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service