Skip to Content
Merck
All Photos(5)

Key Documents

HPA008079

Sigma-Aldrich

Anti-RNF43 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-RING finger protein 43 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200-1:500

immunogen sequence

CVDPWLHQHRTCPLCMFNITEGDSFSQSLGPSRSYQEPGRRLHLIRQHPGHAHYHLPAAYLLGPSRSAVARPPRPGPFLPSQEPGMGPRHHRFPRAAHPRAPGEQQRLAGAQHPYAQGWGLSHLQSTSQH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF43(54894)

Immunogen

RING finger protein 43 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RNF43 (ring finger protein 43) gene encodes a RING-type E3 ubiquitin ligase that functions as a negative regulator of the Wnt signaling pathway. It is involved in the ubiquitination, endocytosis and targeting of Wnt receptor complex components Frizzled receptors to lysosomes. It is localized to the endoplasmic reticulum (ER) and also present in the nuclear envelope. It interacts with HAP95, a chromatin-associated protein, and may regulate cell growth. RNF43 has been implicated in the pathogenesis of colon cancer. It also functions as a tumor suppressor in the intestinal stem cell zone.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71262

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuko Omori et al.
Gastroenterology, 156(3), 647-661 (2018-10-21)
Intraductal papillary mucinous neoplasms (IPMNs) are regarded as precursors of pancreatic ductal adenocarcinomas (PDAs), but little is known about the mechanism of progression. This makes it challenging to assess cancer risk in patients with IPMNs. We investigated associations of IPMNs
Shion Tachibana et al.
Frontiers in oncology, 10, 728-728 (2020-06-26)
Pancreatobiliary tumors frequently contain multiple malignant and precancerous lesions; however, the origin of the driver mutations and the mechanisms that underlie the generation of distinct clones within an organ field remain unclear. Herein, we describe a 76-year-old male suffering from
Haiyang Xie et al.
International journal of clinical and experimental pathology, 8(11), 14995-15000 (2016-01-30)
Our previous study has demonstrated that RNF43 could regulate the cell cycle in a p53-dependent manner in HCC. In this study, we aimed to access whether RNF43 could interact with cell cycle proteins involved in p53 pathway, including pRB, Cyclin
Norma Sainstika Pangestu et al.
Oncology letters, 22(6), 846-846 (2021-11-05)
RING finger protein 43 (RNF43) is a ubiquitin E3 ligase that negatively regulates Wnt/β-catenin signalling. Mutation, inactivation and downregulation of RNF43 in cholangiocarcinoma (CCA) are associated with a less favourable prognosis. Since the functional role of RNF43 in CCA has
Takeyuki Sugiura et al.
Experimental cell research, 314(7), 1519-1528 (2008-03-04)
RNF43 is a recently discovered RING finger protein that is implicated in colon cancer pathogenesis. This protein possesses growth-promoting activity but its mechanism remains unknown. In this study, to gain insight into the biological action of RNF43 we characterized it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service