Skip to Content
Merck
All Photos(7)

Documents

HPA007863

Sigma-Aldrich

Anti-SV2A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Synaptic vesicle glycoprotein 2A antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SDGYYRGEGTQDEEEGGASSDATEGHDEDDEIYEGEYQGIPRAESGGKGERMADGAPLAGVRGGLSDGEGPPGGRGEAQRRKEREELAQQYEAILRECGHGRFQW

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SV2A(9900)

Immunogen

Synaptic vesicle glycoprotein 2A recombinant protein epitope signature tag (PrEST)

Application

Anti-SV2A antibody produced in rabbit has been used for co-immunoprecipitation assay to verify protein-protein interactions.
Anti-SV2A antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

SV2A (Synaptic vesicle glycoprotein 2A) gene encodes a synaptic vesicle protein that functions in signaling at the presynapse. It interacts with and binds to FE65, a binding partner of amyloid precursor protein, and mediates synaptic signal transduction. It is an isoform of SV2, an integral membrane protein that is a part of all synaptic vesicles. SV2A is expressed in the CNS and in endocrine cells. It regulates vesicle function. Knockdown of this isoform and another isoform SV2B in mice, cause severe seizures and postnatal death. In the absence of these isoforms, presynaptic Ca2+ accumulates during consecutive action potentials causing abnormal increases in neurotransmitter release and epilepsy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71567

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oeystein R Brekk et al.
Scientific reports, 8(1), 15207-15207 (2018-10-14)
This report demonstrates insoluble alpha-synuclein (aSYN)+ aggregates in human sporadic Parkinson's disease (PD) midbrain that are linearly correlated with loss of glucocerebrosidase (GCase) activity. To identify early protein-lipid interactions that coincide with loss of lipid homeostasis, an aging study was
Bing Guan et al.
International journal of molecular sciences, 22(23) (2021-12-11)
Neuroendocrine prostate cancer (NEPC) is an aggressive and lethal variant of prostate cancer (PCa), and it remains a diagnostic challenge. Herein we report our findings of using synaptic vesicle glycoprotein 2 isoform A (SV2A) as a promising marker for positron
Hann-Chorng Kuo et al.
European urology, 65(6), 1117-1124 (2014-02-22)
Intradetrusor onabotulinumtoxinA (BoNT-A) injection benefits overactive bladder (OAB) patients, but increased postvoid residual (PVR) urine volume and urinary tract infection (UTI) remain risks. Intravesical instillation of liposomal BoNT-A instead of injection could prevent such adverse events. To evaluate instillation of
Mengfei Xiong et al.
NeuroImage, 277, 120230-120230 (2023-06-25)
Synaptic alterations in certain brain structures are related to cognitive decline in neurodegeneration and in aging. Synaptic loss in many neurodegenerative diseases can be visualized by positron emission tomography (PET) imaging of synaptic vesicle glycoprotein 2A (SV2A). However, the use
Berkley A Lynch et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(26), 9861-9866 (2004-06-24)
Here, we show that the synaptic vesicle protein SV2A is the brain binding site of levetiracetam (LEV), a new antiepileptic drug with a unique activity profile in animal models of seizure and epilepsy. The LEV-binding site is enriched in synaptic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service