Skip to Content
Merck
All Photos(1)

Key Documents

S9381

Sigma-Aldrich

Succinimide

99.1%

Synonym(s):

2,5-Pyrrolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5NO2
CAS Number:
Molecular Weight:
99.09
Beilstein:
108440
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Succinimide,

Assay

99.1%

Quality Level

form

powder or crystals

color

white to off-white

bp

285-290 °C (lit.)

mp

123-125 °C (lit.)

SMILES string

O=C1CCC(=O)N1

InChI

1S/C4H5NO2/c6-3-1-2-4(7)5-3/h1-2H2,(H,5,6,7)

InChI key

KZNICNPSHKQLFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Rennison et al.
Bioorganic & medicinal chemistry, 15(8), 2963-2974 (2007-02-27)
Norbormide [5-(alpha-hydroxy-alpha-2-pyridylbenzyl)-7-(alpha-2-pyridylbenzylidene)-5-norbornene-2,3-dicarboximide] (NRB, 1), an existing but infrequently used rodenticide, is known to be uniquely toxic to rats but relatively harmless to other rodents and mammals. A series of NRB-related analogues were prepared to investigate the structural features responsible for
Nada Perisic-Janjic et al.
Journal of pharmaceutical and biomedical analysis, 72, 65-73 (2012-11-14)
Reversed-phase thin-layer chromatographic (RP TLC) retention coefficients for a newly designed series of N-phenyl-3-methyl succinimide derivatives, of a rationally expected anticonvusant activity, were determined as parameters of their lipophilicity. Basic pharmacokinetic descriptors of the agents were calculated in silico with
Hyun-Ok Song et al.
PloS one, 7(11), e48459-e48459 (2012-11-08)
Rapid in-field diagnosis is very important to prevent the outbreak of various infectious and contagious diseases. Highly sensitive and quantitative detection of diseases can be performed using fluorescent immunochemical assay with specific antigen-antibody binding and a good quality fluorophore. This
Wataru Kamimura et al.
Journal of biomaterials science. Polymer edition, 23(5), 609-628 (2011-02-12)
We have developed a scaffold material consisting of a covalently-bonded structure of alginate and atelocollagen (AtCol). Addition of calcium ions caused the material to form a hydrogel (alginate-modified AtCol gel). The condition of the alginate-modified AtCol gel could be controlled
Zhiwei Lin et al.
Physical chemistry chemical physics : PCCP, 14(30), 10445-10454 (2012-05-05)
Dual-frequency relaxation-assisted two-dimensional infrared (RA 2DIR) spectroscopy was used to investigate energy transport in polyethylene glycol (PEG) oligomers of different length, having 0, 4, 8, and 12 repeating units and end-labeled with azido and succinimide ester moieties (azPEGn). The energy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service