Skip to Content
Merck
All Photos(3)

Key Documents

S0520

Sigma-Aldrich

Selenocystamine dihydrochloride

>98% (TLC), suitable for ligand binding assays

Synonym(s):

2,2′-diselenobis-Ethanamine hydrochloride (1:2)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H12N2Se2 · 2HCl
CAS Number:
Molecular Weight:
318.99
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

Selenocystamine dihydrochloride, powder

Assay

>98% (TLC)

Quality Level

form

powder

technique(s)

ligand binding assay: suitable

color

yellow to orange

solubility

H2O: soluble, clear to slightly hazy

SMILES string

NCC[Se][Se]CCN.[H]Cl.[H]Cl

InChI

1S/C4H12N2Se2.ClH/c5-1-3-7-8-4-2-6;/h1-6H2;1H

InChI key

PKRYDZYGNSZBHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Selenocystamine dihydrochloride has a potential to block the activity of influenza A and B virus associated RNA-dependent RNA polymerase enzyme.[1]

Application

Selenocystamine dihydrochloride has been used to determine its effect on PP2A phosphatase activity in vitro.[2] It has also been used as a catalyst for a disulfide-cleaving reagent, dithiothreitol (DTT).[3]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conquest of viral diseases: a topical review of drugs and vaccines, 1 (1985)
Di Ding et al.
Biochemical and biophysical research communications, 361(3), 629-633 (2007-08-04)
Two distinct stereospecific methionine sulfoxide reductases (Msr), MsrA and MsrB reduce the oxidized methionine (Met), methionine sulfoxide [Met(O)], back to Met. In this report, we examined the reducing systems required for the activities of two chloroplastic MsrB enzymes (NtMsrB1 and
Y Hou et al.
Biochemical and biophysical research communications, 228(1), 88-93 (1996-11-01)
Seleno compounds such as selenocystamine and seleno-D, L-cystine were found to catalyze the decomposition of S-nitrosothiols (e.g. S-nitroso-glutathione and S-nitroso-N-acetyl-D, L-penicillamine) in the presence of different thiols (e.g. glutathione, N-acetyl-D-penicillamine and 2-mercaptoethanol), and liberate nitric oxide. It was also found
Sodium selenate specifically activates PP2A phosphatase, dephosphorylates tau and reverses memory deficits in an Alzheimer?s disease model
Corcoran NM, et al.
Journal of Clinical Neuroscience : Official Journal of the Neurosurgical Society of Australasia, 17(8), 1025-1033 (2010)
Transferrin serves as a mediator to deliver organometallic ruthenium (II) anticancer complexes into cells
Guo W, et al.
Inorganic Chemistry, 52(9), 5328-5338 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service