Skip to Content
Merck
All Photos(1)

Key Documents

S8140

Sigma-Aldrich

SSPE Buffer

for Northern and Southern blotting, powder blend

Synonym(s):

Saline-Sodium Phosphate-EDTA Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319
NACRES:
NA.25

grade

for molecular biology

Quality Level

form

powder blend

pH

7.2-7.6 (20 × in water, high purity, ambient)

solubility

water, high purity: 20 ×, clear, colorless

foreign activity

DNAse, none detected
Endonuclease, none detected
Exonuclease, none detected
NICKase, none detected
RNAse, none detected

storage temp.

room temp

General description

SSPE Buffer 20x powder is a standard reagent in Southern and Northern hybridization procedures. This powder should be reconstituted to the indicated volume with molecular biology grade water before use. The resulting 20x liquid can be used straight or diluted as needed.

Application


  • Influence of Saline Buffers over the Stability of High-Annealed Gold Nanoparticles Formed on Coverslips for Biological and Chemosensing Applications.: This study explores the role of saline buffers in maintaining the stability of high-annealed gold nanoparticles used in biological and chemosensing applications. The findings highlight the potential for enhanced sensor performance in detecting biological molecules (Zhou & Ionescu, 2020).

  • Quantification of Surface Etching by Common Buffers and Implications on the Accuracy of Label-Free Biological Assays.: This research assesses the impact of various buffers, including SSPE buffer, on the surface etching of substrates used in label-free biological assays. The results indicate that buffer-induced surface changes can significantly affect assay accuracy (Ahn et al., 2012).

Features and Benefits

  • Easy to reconstitute powder
  • Multi-purpose use in hybridizations

Components

Final SSPE Buffer 20x contains 0.02M EDTA and 2.98M NaCl in 0.2M phosphate buffer (pH 7.4).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microfluidic-based enzymatic on-chip labeling of miRNAs
Sonja Vorwek
New Biotechnology (2008)
3'-Minor groove binder-DNA probes increase sequence specificity at PCR extension temperatures
Igor V. Kutyavin
Nucleic Acids Research, 28(2) (2000)

Protocols

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service