Skip to Content
Merck
All Photos(1)

Key Documents

D0428

Sigma-Aldrich

1 kb DNA Ladder

for DNA electrophoresis

Synonym(s):

1kb gel ladder, 1kb gel marker, 1kb ladder for gel electrophoresis, DNA gel marker, agarose gel electrophoresis ladder, agarose gel electrophoresis marker

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105335
NACRES:
NA.25

Quality Level

form

liquid

suitability

suitable for electrophoresis (DNA)

storage temp.

−20°C

General description

Sigma′s 1 kb Ladder contains 11 fragments consisting of 500 bp repeats from 0.5 to 3 kb, 1 kb repeats from 3 to 6 kb, and 2 kb repeats from 6 to 10 kb. In nucleic acid gel electrophoresis, five μl of the marker should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel. Suitable for use in Northern and Southern blotting. All DNA markers can be stained with ethidium bromide, SYBR® Green, and Nancy-520.

Application

1 kb DNA Ladder has been used as a molecular marker to determine the molecular weight and size of double stranded DNA during gel electrophoresis.

Components

Sigma′s 1kb Ladder is provided in a solution of 10 mM Tris-HCl (pH 8.0), with 1.0 mM EDTA.

Other Notes

For optimal resolution, the recommended agarose gel concentration is 0.75%.

Legal Information

SYBR is a registered trademark of Life Technologies

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Zhou et al.
Journal of experimental botany, 59(10), 2803-2813 (2008-06-03)
The turnip crinkle virus-based vector TCV-GFP Delta CP had been devised previously to study cell-to-cell and long-distance spread of virus-induced RNA silencing. TCV-GFP Delta CP, which had been constructed by replacing the coat protein (CP) gene with a green fluorescent
Lidia Fuentes et al.
Journal of inorganic biochemistry, 203, 110875-110875 (2019-11-11)
One mononuclear and another dinuclear Pd(II) complexes bearing a α-N-heterocyclic thiosemicarbazone ligand have been synthesized, fully characterized and studied as biological agents. In both complexes, the palladium center is coordinated to 3,5-diacetyl-1,2,4-triazol bis(N4,N4-dimethylthiosemicarbazone) via three sites (N, N and S).
W S Choe et al.
Biotechnology progress, 17(6), 1107-1113 (2001-12-12)
The direct chemical extraction of recombinant L1 protein (the major capsid protein of human papillomavirus type 16) from the cytoplasm of E. coli HMS174(DE3) has recently been demonstrated at high cell density (to OD(600) = 160) without the use of
Luca Cocolin et al.
Applied and environmental microbiology, 70(3), 1347-1355 (2004-03-10)
In this paper we describe the development of a PCR protocol to specifically detect Brettanomyces bruxellensis and B. anomalus. Primers DB90F and DB394R, targeting the D1-D2 loop of the 26S rRNA gene, were able to produce amplicons only when the
Ana I Matesanz et al.
Chembiochem : a European journal of chemical biology, 21(8), 1226-1232 (2019-11-21)
The synthesis and characterization of three new platinum complexes, with 3,5-diacetyl-1,2,4-triazole bis(4-N-isopropylthiosemicarbazone) as a ligand, are reported. The specific conditions under which solvent coordination takes place are reported and the X-ray structure of the complex with one solvent molecule of

Articles

Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.

Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.

Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.

Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service