Skip to Content
Merck
All Photos(1)

Key Documents

E1250

Sigma-Aldrich

Elastase from porcine pancreas

Type I, ≥4.0 units/mg protein

Synonym(s):

Elastase from hog pancreas, Pancreatopeptidase E

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

Porcine pancreas

Quality Level

type

Type I

form

suspension

specific activity

≥4.0 units/mg protein

mol wt

25.9 kDa

contains

0.1% thymol

concentration

0.5-15.0 mg/mL in water

foreign activity

trypsin ≤50 BAEE units/mg protein

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Elastase from porcine pancreas has been used in a study to investigate the design, synthesis and evaluation of biomimetic affinity ligands for elastases. Elastase from porcine pancreas has also been used in a study to investigate the purification and partial characterization of the pancreatic proteolytic enzymes trypsin, chymotrypsin, and elastase.
Elastase from porcine pancreas has been used:
  • to induce abdominal aortic aneurysm (AAA)
  • to study the impact of indoleamine 2-3 dioxygenase 1 (IDO) in mice[1]
  • to digest aortas for aortic smooth muscle cells (SMC) isolation[2]
  • as a positive control of proteolytic digestion[3]

The enzyme from Sigma has been used in the development of elastase-perfused animal model . This study determined if tobacco exposure could lower the threshold of aortic injury necessary for AAA (abdominal aortic aneurysm) development.[4] It has also been used during the isolation of type II pneumocytes from human lungs.[5]

Biochem/physiol Actions

Elastase hydrolyses elastin, the specific protein of elastic fibers, and digests hemoglobin, casein and fibrin.
Elastase is a single polypeptide chain of 240 amino acid residues and contains four disulfide bridges. The molecular mass is approximately 25.9 kDa. The enzyme is synthesized as an inactive zymogen, proelastase, which is converted to the active form by limited proteolysis at the N-terminal by trypsin.[6] It is a serine protease with broad specificity. It cleaves protein at the carboxyl side of small hydrophobic amino acids such as Ile, Gly, Ala, Ser, Val, and Leu. The enzyme also hydrolyzes amides and esters such as N-Benzoyl-L-alanine methyl ester.[6][7] The pH optimum is found to be 8.0-8.5. It does not require any activator, but it is inhibited by diisopropyl fluorophosphate, phenylmethanesulfonyl fluoride, α2-macroglobulin, α1-antitrypsin, sulfonyl fluorides and p-dinitrophenyl diethylphosphate and high salt concentrations. It is extensively used in tissue and cell dissociation procedures. Elastase is effective in the isolation of Type II lung cells.

Packaging

Package size based on protein content

Unit Definition

One unit will hydrolyze 1.0 μmole of N-succinyl-L-Ala-Ala-Ala-p-nitroanilide per min, pH 8.0 at 25 °C.

Preparation Note

2× crystallized

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liren Duan et al.
Frontiers in physiology, 11, 866-866 (2020-08-09)
Aim: Smoking is a major risk factor for abdominal aortic aneurysm (AAA). Among the components of smoke, nicotine is known to exert pro-atherosclerotic, prothrombotic, and proangiogenic effects on vascular smooth muscle cells (VSMCs). The current study was designed to investigate
Indoleamine 2 3-dioxygenase knockout limits angiotensin II-induced aneurysm in low density lipoprotein receptor-deficient mice fed with high fat diet
Metghalchi S, et al.
PLoS ONE, 13(3), e0193737-e0193737 (2018)
Demetrios Moris et al.
Current vascular pharmacology, 16(2), 168-178 (2017-02-06)
Abdominal aortic aneurysm (AAA) formation is associated with by inflammation and matrix degradation. This study tested the hypothesis that calprotectin, a novel biomarker for inflammation, as well as established biomarkers such as C-reactive protein (CRP) and matrix metalloproteinase- 9 (MMP-9)
A C Cunningham et al.
Journal of cell science, 107 ( Pt 2), 443-449 (1994-02-01)
Highly purified populations of alveolar epithelial cells (type II pneumocytes) were isolated from human lung specimens. These cells were characterised histochemically, by demonstrating the presence of intracellular alkaline phosphatase, and morphologically, by electron microscopic demonstration of lamellar bodies and microvilli.
Michel P Bergoeing et al.
Journal of vascular surgery, 45(6), 1217-1227 (2007-04-03)
The development of abdominal aortic aneurysms (AAA) is presumed to result from multiple genetic and environmental factors, with exposure to tobacco smoke the single largest known factor predisposing to aneurysm growth. We have attempted to adapt the elastase-perfused animal model

Protocols

Enzymatic Assay of Elastase (EC 3.4.21.36), a continuous spectrophotometric rate determination.

Enzymatic Assay of Elastase (EC 3.4.21.36), a continuous spectrophotometric rate determination.

Enzymatic Assay of Elastase (EC 3.4.21.36), a continuous spectrophotometric rate determination.

Enzymatic Assay of Elastase (EC 3.4.21.36), a continuous spectrophotometric rate determination.

Chromatograms

application for HPLC

Questions

  1. What is the difference between Type I and Type IV Elastase?

    1 answer
    1. The major differences between the different types of Elastase are as follows:

      E1250: Type I, aqueous suspension, >4 units/mg protein, <50 units/mg protein Trypsin. Contains Thymol as an antimicrobial, which is toxic to cells.

      E6883: Type IIA, discontinued

      E0127: Type III, lyophilized powder, >4 units/mg protein, <200 units/mg protein Trypsin.

      E0258: Type IV, lyophilized powder, >4 units/mg protein, <50 units/mg protein Trypsin, further purification of E0127.

      E7885: Bioreagent, lyophilized powder, suitability tested for use in cell culture, >4 units/mg protein, <50 units/mg protein Trypsin.

      45125: Lyophilized powder, >6.4 units/mg protein, not tested for Trypsin impurity.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service