Skip to Content
Merck
All Photos(1)

Key Documents

46842

Supelco

Sulfamethizole

VETRANAL®, analytical standard

Synonym(s):

4-Amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfonamide, N1-(5-Methyl-1,3,4-thiadiazol-2-yl)sulfanilamide, Sulfamethylthiadiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N4O2S2
CAS Number:
Molecular Weight:
270.33
Beilstein:
255002
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

Cc1nnc(NS(=O)(=O)c2ccc(N)cc2)s1

InChI

1S/C9H10N4O2S2/c1-6-11-12-9(16-6)13-17(14,15)8-4-2-7(10)3-5-8/h2-5H,10H2,1H3,(H,12,13)

InChI key

VACCAVUAMIDAGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Robert R Muder
Seminars in respiratory and critical care medicine, 28(6), 672-677 (2007-12-21)
Stenotrophomonas (Xanthomonas) maltophilia is a nonfermentative, gram-negative bacillus that is widely distributed in natural and human-made environments. In the nonhospital setting it is an uncommon pathogen, typically causing soft tissue infection of contaminated wounds. In the hospital setting, particularly among
Ali Md Ahad et al.
Bioorganic & medicinal chemistry, 19(6), 2046-2054 (2011-03-01)
Disruption of the phosphatidylinositol 3-kinase/AKT signaling pathway can lead to apoptosis in cancer cells. Previously we identified a lead sulfonamide that selectively bound to the pleckstrin homology (PH) domain of AKT and induced apoptosis when present at low micromolar concentrations.
Tony L Yaksh et al.
Anesthesiology, 108(5), 938-949 (2008-04-24)
Intrathecal N-methyl-d-aspartate antagonists have antihyperalgesic efficacy. The authors examined toxicity in a canine model of chronic lumbar intrathecal infusion. Dogs (10-16 kg) were prepared with lumbar intrathecal catheters connected to vest-mounted pumps (100 microl/h). In phase 1, stepwise incrementations in
Mark R Sannes et al.
The Journal of infectious diseases, 197(3), 430-434 (2008-01-11)
To determine whether poultry contact/consumption predicts colonization with antimicrobial-resistant Escherichia coli, 567 newly hospitalized patients and 100 vegetarians were assessed microbiologically and epidemiologically. Multivariable analysis showed that poultry contact/consumption, other dietary habits, and antimicrobial use did not significantly predict resistance.
K Koligi et al.
Der Internist, 52(7), 884-888 (2010-11-04)
Whipple's disease is a rare, chronic infection caused by Tropheryma whipplei, an ubiquitary gram positive bacterium. The disease is associated with a high mortality in absence of an antibiotic treatment. The disease can be detected in affected tissues and body

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service