Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sudan III
Synonym(s):
1-[4-(Phenylazo)phenylazo]-2-naphthol, Cerasin Red, Fat Ponceau G, Fat Soluble Sudan, Scarlet B, Solvent Red 23, Sudan G, Sudan Red BK, Tony Red
Empirical Formula (Hill Notation):
C
22
H
16
N
4
O
CAS Number:
85-86-9
Molecular Weight:
352.39
EC Number:
201-638-4
Products
Product Category
stains (2)
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S4131
Sudan III,
Dye content, ≥85%, technical grade, powder
Expand
View Pricing
S4136
Sudan III,
Dye content, ≥80%, certified by the Biological Stain Commission, powder
Expand
View Pricing
68562
Sudan III,
analytical standard
Expand
View Pricing