Skip to Content
Merck
All Photos(3)

Key Documents

S3147

Sigma-Aldrich

5-Sulfosalicylic acid dihydrate

suitable for electrophoresis, ≥99%

Synonym(s):

2-Hydroxy-5-sulfobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO3SC6H3-2-(OH)CO2H·2H2O
CAS Number:
Molecular Weight:
254.21
Beilstein:
650741
EC Number:
MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:
NACRES:
NB.22

Assay

≥99%

form

powder or crystals

technique(s)

electrophoresis: suitable

impurities

≤0.02% Insoluble matter
≤0.05% Salicylic Acid

mp

105-110 °C (lit.)

solubility

water: soluble 100 mg/mL, clear, colorless

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.035%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.002%

SMILES string

[H]O[H].[H]O[H].OC(=O)c1cc(ccc1O)S(O)(=O)=O

InChI

1S/C7H6O6S.2H2O/c8-6-2-1-4(14(11,12)13)3-5(6)7(9)10;;/h1-3,8H,(H,9,10)(H,11,12,13);2*1H2

InChI key

BHDKTFQBRFWJKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Sulfosalicylic acid is a strong acid capable of protonating water. It forms a new theophylline complex, which was investigated by X-ray photoelectron spectroscopy (XPS). It forms 1:1 proton-transfer compounds with the ortho-substituted monocyclic heteroaromatic Lewis bases. Their crystal structures have been studied.

Application

5-Sulfosalicylic acid (3.5%) in 12% trichloroacetic acid is commonly used as a fixing solution for proteins in agarose and polyacrylamide gels.
5-Sulfosalicylic acid dihydrate may be employed for the quantification of doxorubicin derived from PEGylated liposomal doxorubicin (Doxil) and its major metabolite in human plasma by HPLC. It may be used in the preparation of poly[[tris(di-2-pyridylamine)tris([μ]-5-sulfonatosalicylato)tricopper(II)] trihydrate].

Analysis Note

Tested for use in fixing solutions for PAGE, SDS-PAGE and IEF.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David L Chin et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 779(2), 259-269 (2002-10-04)
A high-performance liquid chromatographic method was developed for the quantification of doxorubicin derived from PEGylated liposomal doxorubicin (Doxil) and its major metabolite in human plasma. This method utilizes Triton X-100 to disperse the liposome, followed by a protein precipitation step
Joanna S Stevens et al.
The journal of physical chemistry. B, 114(44), 13961-13969 (2010-10-22)
Recent studies suggested that X-ray photoelectron spectroscopy (XPS) sensitively determines the protonation state of nitrogen functional groups in the solid state, providing a means for distinguishing between co-crystals and salts of organic compounds. Here we describe how a new theophylline
Megan A Dickson et al.
Reproductive biology and endocrinology : RB&E, 18(1), 49-49 (2020-05-16)
Cigarette smokers have a reduced risk of developing preeclampsia, possibly attributed to an increase in carbon monoxide (CO) levels. Carbon monoxide is a gasotransmitter that has been implicated in maintaining vascular tone, increasing angiogenesis, and reducing inflammation and apoptosis at
Hydrogen bonding in proton-transfer compounds of 5-sulfosalicylic acid with ortho-substituted monocyclic heteroaromatic Lewis bases.
Smith G, et al.
Journal of Chemical Crystallography, 36(12), 841-849 (2006)
Poly [[tris (di-2-pyridylamine) tris (μ-5-sulfonatosalicylato) tricopper (II)] trihydrate].
Fan SR and Zhu LG.
Acta Crystallographica Section E, Structure Reports Online, 61(1), m174-m176 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service