Skip to Content
Merck
All Photos(1)

Documents

S1764

Sigma-Aldrich

Complement sera human

lyophilized powder

Synonym(s):

Human Serum Complement

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human

Quality Level

form

lyophilized powder

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

virus, tested

shipped in

dry ice

storage temp.

−20°C

General description

Complement serum is prepared from pooled human plasma and lyophilized from the amount of serum indicated on the label.

Application

Complement sera human has been used:
  • to incubate DLD-1 or SW-620 cells to quantify C5a release
  • in multiplex bead antibody-binding assay to assay C3d
  • in complement-dependent cytotoxicity assay

Complement sera human has been used:
  • to incubate DLD-1 or SW-620 cells to quantify C5a release
  • in multiplex bead antibody-binding assay to assay C3d
  • in complement-dependent cytotoxicity assay

The activity of complement sera can be determined by hemolytic assays in vitro.

Physical form

Lyophilized powder from indicated amount of serum

Analysis Note

Hemolytic titer (CH50 units per ml) is determined by method of Kabat and Mayer. Actual titer given on label.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David A Spencer et al.
Nature communications, 13(1), 662-662 (2022-02-05)
Increasingly, antibodies are being used to treat and prevent viral infections. In the context of HIV, efficacy is primarily attributed to dose-dependent neutralization potency and to a lesser extent Fc-mediated effector functions. It remains unclear whether augmenting effector functions of
Chiaki Kawabata et al.
Biological & pharmaceutical bulletin, 44(2), 219-224 (2021-02-02)
Baculovirus vectors (BVs) are safely able to transduce foreign genes and express them in mammalian cells. However, the transduction activity of BVs is strongly reduced by the attack of serum complement, which is one of the major obstacles in the
Helen Kotanides et al.
Cancer immunology research, 8(10), 1300-1310 (2020-09-03)
The programmed cell death protein 1 receptor (PD-1) and programmed death ligand 1 (PD-L1) coinhibitory pathway suppresses T-cell-mediated immunity. We hypothesized that cotargeting of PD-1 and PD-L1 with a bispecific antibody molecule could provide an alternative therapeutic approach, with enhanced
Savannah E Butler et al.
Frontiers in immunology, 11, 618685-618685 (2021-02-16)
Understanding humoral immune responses to SARS-CoV-2 infection will play a critical role in the development of vaccines and antibody-based interventions. We report systemic and mucosal antibody responses in convalescent individuals who experienced varying severity of disease. Whereas assessment of neutralization
Anaïs Thiriard et al.
Frontiers in immunology, 14, 1107156-1107156 (2023-04-04)
To comprehensively analyze the quality of the antibody response between children with Multisystem inflammatory syndrome (MIS-C) and age-matched controls at one month after SARS-CoV-2 exposure, and infected in the same time-period. Serum from 20 MIS-C children at admission, and 14

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service