Skip to Content
Merck
All Photos(1)

Key Documents

H4522

Sigma-Aldrich

Human Serum

from human male AB plasma, USA origin, sterile-filtered

Synonym(s):

human plasma, human sera, human serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71
biological source:
human male plasma (Type AB)
origin:
USA origin
sterility:
sterile-filtered
shipped in:
dry ice
Pricing and availability is not currently available.

biological source

human male plasma (Type AB)

Quality Level

sterility

sterile-filtered

composition

hemoglobin, ≤30 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable
cell culture | stem cell: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Human AB serum is used in tissue engineering, transplantation and cell therapy applications for the expansion of mesenchymal stem cells (MSC) from adipose tissue or mesenchymal stromal cells from human bone marrow; for standardized limbal epithelial stem cell graft generation and transplantation; for ex vivo expansion of NK cells from peripheral blood in Hematopoeitic Stem Cell Expansion Medium; and for upgrading pretransplant human islet culture technology.
Human Serum has been used: as a component of phosphate-buffered saline with Tween® detergent (PBST) in bridging enzyme-linked immunosorbent assay (ELISA), as a mock biological sample for assessing the performance of paper-based ELISA assay with complex biological samples, as a diluent to create a dilution series of the baseline sample for the generation of standard curve for NISTmAb spiked into serum, as well as for multiple myeloma (MM) patient sample MM2232

Biochem/physiol Actions

Human AB serum is typically obtained by allowing whole blood to clot naturally, which is known as Off-The-Clot (OTC) serum. Human serum obtained from clotted whole blood is preferred as it usually contains higher levels and a more diverse range of growth factors for optimal cell growth. Human serum from blood-type AB donors lacks antibodies specific for both A and B blood-type antigens. This makes it a universal donor serum that is suitable for various cell culture types including human stem cell lines.

Packaging

Packaging may vary for different batches due to short supply of the bottles used to pack this product. The presence or lack of shrink wrap around the cap of the bottle doesn’t impact product integrity/sterility of the product.

Warning

Each donor is tested for and found non-reactive for Hepatitis B & C and non-reactive for Human Immunodeficiency Virus (HIV) antibody by ELISA. Nevertheless, products of human origin should be considered potentially infectious and handled accordingly.

Analysis Note

Endotoxin tested

Other Notes

It is not unusual for our human serum products to be cloudy. This product is from pooled serum and cloudiness indicates that lipids are present in the serum. No clear, lipid-free human serum products are available.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason P Wong et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(33), 16541-16550 (2019-07-28)
Non-Hodgkin lymphomas (NHLs) make up the majority of lymphoma diagnoses and represent a very diverse set of malignancies. We sought to identify kinases uniquely up-regulated in different NHL subtypes. Using multiplexed inhibitor bead-mass spectrometry (MIB/MS), we found Tyro3 was uniquely
Michelle Baker et al.
Scientific reports, 7(1), 8329-8329 (2017-08-23)
In worldwide conditions of increasingly antibiotic-resistant hospital infections, it is important to research alternative therapies. Bdellovibrio bacteriovorus bacteria naturally prey on Gram-negative pathogens, including antibiotic-resistant strains and so B. bacteriovorus have been proposed as "living antibiotics" to combat antimicrobially-resistant pathogens.
Lawrence Fong et al.
Cancer discovery, 10(1), 40-53 (2019-11-17)
Adenosine mediates immunosuppression within the tumor microenvironment through triggering adenosine 2A receptors (A2AR) on immune cells. To determine whether this pathway could be targeted as an immunotherapy, we performed a phase I clinical trial with a small-molecule A2AR antagonist. We
Iria Gomez-Tourino et al.
Nature communications, 8(1), 1792-1792 (2017-11-28)
Defects in T cell receptor (TCR) repertoire are proposed to predispose to autoimmunity. Here we show, by analyzing >2 × 10
Marta Monguió-Tortajada et al.
Cell death & disease, 9(5), 499-499 (2018-05-03)
TLR sensing of pathogens triggers monocyte activation to initiate the host innate immune response to infection. Monocytes can dynamically adapt to different TLR agonists inducing different patterns of inflammatory response, and the sequence of exposure to TLRs can dramatically modulate

Articles

Learn about the impact of filtration on hematopoietic cell culture media containing vital T cell factors and other media components.

Learn about the impact of filtration on hematopoietic cell culture media containing vital T cell factors and other media components.

Learn about the impact of filtration on hematopoietic cell culture media containing vital T cell factors and other media components.

Learn about the impact of filtration on hematopoietic cell culture media containing vital T cell factors and other media components.

See All

Questions

1–10 of 20 Questions  
  1. What is the antibodies concentration in Product H4522, human serum?

    1 answer
    1. The immunoglobulin concentration of this product is not measured. This is a pooled product from adult AB negative males. Based on information published in the Geigy Scientific Tables, the average immunoglobulin levels for adult males, ages 20 – 70, are as follows:
      IgG
      10.69 – 12.04 g./L
      IgA
      1.32 – 2.34 g/L
      IgM (not age dependent)
      0.77 g/L

      Helpful?

  2. Il prodotto è stato testato per negatività a HSV?

    1 answer
    1. Each donor is tested for and found non-reactive to Hepatitis B & C and non-reactive to Human Immunodeficiency Virus (HIV) antibody by ELISA. HSV is not on the list of agents that are tested. Nevertheless, products of human origin should be considered potentially infectious and handled accordingly. See the link below to review a sample or lot specific Certificate of Analysis for this product: https://www.sigmaaldrich.com/product/sigma/i4506#product-documentation

      Helpful?

  3. If this is "serum" derived from "plasma" the plasma would have contained anti-coagulant. To turn the plasma into serum it has to be clotted.  Has this product  been clotted to remove the fibrin? Or is it still in fact plasma?

    1 answer
    1. The product has been clotted. Blood is collected and treated with citrate (anticoagulant) which inhibits clotting initially. The blood is then centrifuged. When the spin is complete, layers will appear which allow a differentiation between the cells and the plasma. The plasma is removed and calcium is added to activate the clotting cascade. The preparation is then centrifuged again and the result is a large clot which is pelleted to the bottom of the tube with a serum layer on top.

      Helpful?

  4. Are Human Serum products H4522 and H6914 suitable for complement assays? Is there testing available for complement levels or activity?

    1 answer
    1. Human Serum products H4522 and H6914 may not be suitable for complement assays when stored at -20°C due to potential degradation of complement activity. For complement assays, alternatives like S1764 (Complement sera human) and C9473 (Complement Serum Standard human) are recommended. It is advised to review the Certificates of Analysis (CoAs) for these products to ensure they meet the specific requirements of your complement assay

      Helpful?

  5. Which human serum is recommend for a nucleic acid stability assay, specifically for incubating siRNA in serum to represent bloodstream kinetics?

    1 answer
    1. The use of human serum for a nucleic acid stability assay has not been performed as it is not part of our lab's product characterization focus. However, the citation titled "Improving gene silencing of siRNAs via tricyclo-DNA modification, PMID: 21687522" suggest that product H4522 may be suitable for this application. It is recommended to refer to the publication for additional insights.

      Helpful?

  6. What are the differences between serum and plasma-derived serum?

    1 answer
    1. Whole blood serum is obtained by allowing natural clotting followed by filtration,lacking fibrinogen. Plasma-derived serum is obtained from non-clotted whole blood through centrifugation.

      Helpful?

  7. What Biosafety Level would be suitable for human serum such as H4522, H5667, and H6914?

    1 answer
    1. Human origin products from normal donors should be used at Biosafety Level 2 (BSL-2), which is suitable for handling moderate-risk agents causing human disease. Thus, treat these products as potentially infectious and handle accordingly.

      Helpful?

  8. what does this do

    1 answer
    1. Human AB serum is added as a supplement to culture media at a concentration of 2-10% to provide a mixture of nutritional, hormonal, growth, and attachment factors. The serum also acts as a buffer to the cell culture system against a variety of disruptions to cell growth and toxic effects such as pH change, proteolytic activity, or the presence of heavy metals or endotoxin.

      Please see the link below for more information:
      https://www.sigmaaldrich.com/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/mammalian-cell-culture/serum-cell-tissue-faq

      Helpful?

  9. Is product H4522 obtained using Institutional Review Board (IRB)-approved consent forms and protocols?

    1 answer
    1. According to the supplier:
      Each donor unit used in the preparation of this material was tested and found negative for HBsAG, HBV-NAT, HIV-1/2, HCV, HCV-NAT, HIV-1 NAT, and Syphilis per FDA regulations. The human plasma was collected from male donors' blood type AB in FDA licensed commercial donor establishments located in the USA in accordance with ethical standards. Each donor signed an informed consent form prior to collection, which is traceable by the unique identifiable number to maintain anonymity. A quality assurance system is in place to ensure traceability to individual donor. Documentation to this effect is available upon request.

      Helpful?

  10. Could you please confirm if the serum contains normal levels of human complement components?

    1 answer
    1. This product has not been treated to remove or inactivate complement components, however no testing has been performed to confirm these levels. It is a pooled product derived from 50 - 70 donors per lot. Any active complement be expected to meet 'normal levels.

      Should complement testing be a requirement, please see product S1764. This is a lyophilized human serum product tested for CH50 activity. See the links below to review a sample Certificate of Analysis and the product datasheet:

      S1764 Certificate of Analysis
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/S1764-1MLSLCQ7524.pdf

      S1764 Datasheet
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/408/363/s1764dat.pdf

      Helpful?

1–10 of 20 Questions  

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service