Skip to Content
Merck
All Photos(1)

Key Documents

C2910

Sigma-Aldrich

Complement C3 from human serum

≥3000 C3H50 units/mg (using C3 deficient serum)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human

Quality Level

Assay

≥85% (SDS-GE)

form

solution

specific activity

≥3000 C3H50 units/mg (using C3 deficient serum)

mol wt

185 kDa (disulfide-linked heterodimer)

technique(s)

activity assay: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... C3(718)

Looking for similar products? Visit Product Comparison Guide

Application

Complement C3 is important in complement activation through the classical and alternative pathways. Complement C3 has been used in studies of bacterial defense against the complement system. Research on streptococcus infection has shown that Streptococcal pyrogenic exotoxin B (SPE B), a cysteine protease, can prevent phagocytic activity via cleavage of the C3 and impairing the pathways of the compliment system. C3 deficiency can result in recurrent infections and immune complex disorders. Recent research has shown that inherited C3 deficiency can occur either through homozygous mutations or through compound heterozygous mutations,

Biochem/physiol Actions

Complement C3 is the third and most abundant component of the complement pathway. It plays a central role in complement activation, being involved in both the classical and alternative pathways. C3 or its proteolytic fragments mediate many biological functions such as opsonization and anaphylatoxin activities. Pro-C3 is synthesized as a large single chain of 185 kDa and is processed to a disulfide-linked heterodimer consisting of an α-chain (115 kDa) and a β-chain (70 kDa) in the plasma. C3 is cleaved by the C3 convertases in either pathway (C4b2a, C3iBb, C3bBb) to C3b (176 kDa) comprised of the large C-terminal portion of the α-chain and the β-chain. C3b interacts with other complement components to initiate the amplification cascade.
Complement C3 is the third and most abundant component of the complement pathway. It plays a central role in complement activation, being involved in both the classical and alternative pathways. C3 or its proteolytic fragments mediate many biological functions such as opsonization and anaphylatoxin activities. Pro-C3 is synthesized as a large single chain of 185 kDa and is processed to a disulfide-linked heterodimer consisting of an α-chain (115 kDa) and a β-chain (70 kDa) in the plasma. C3 is cleaved by the C3 convertases in either pathway (C4b2a, C3iBb, C3bBb) to C3b (176 kDa) comprised of the large C-terminal portion of the α-chain and the β-chain. C3b interacts with other complement components to initiate the amplification cascade. Studies of C3 function have been facilitated by experimentally induced C3 deficiency in animals by injection of cobra venom factor and more recently with gene ablation technology.

Quality

Functionally pure by a hemolytic assay using deficient sera.

Physical form

Supplied as a solution in PBS, pH 7.2.

Other Notes

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chih-Feng Kuo et al.
Infection and immunity, 76(3), 1163-1169 (2008-01-05)
Streptococcal pyrogenic exotoxin B (SPE B), a cysteine protease, is an important virulence factor in group A streptococcus (GAS) infection. The inhibition of phagocytic activity by SPE B may help prevent bacteria from being ingested. In this study, we examined
Bin Li et al.
Toxicological sciences : an official journal of the Society of Toxicology, 107(1), 144-155 (2008-10-22)
The expectation from the literature is that organophosphorus (OP) agents bind to proteins that have an active site serine. However, transferrin, a protein with no active site serine, was covalently modified in vitro by 0.5mM 10-fluoroethoxyphosphinyl-N-biotinamido pentyldecanamide, chlorpyrifos oxon, diisopropylfluorophosphate
Yutaka Terao et al.
The Journal of biological chemistry, 283(10), 6253-6260 (2007-12-28)
A relative lack of neutrophils around Streptococcus pyogenes is observed in streptococcal toxic shock syndrome (STSS). Because the bacteria spread rapidly into various organs in STSS, we speculated that S. pyogenes is equipped with molecules to evade the host innate
P Randhawa et al.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 15(4), 1014-1020 (2015-03-05)
Neutralizing antibodies (NAbs) form the basis of immunotherapeutic strategies against many important human viral infections. Accordingly, we studied the prevalence, titer, genotype-specificity, and mechanism of action of anti-polyomavirus BK (BKV) NAbs in commercially available human immune globulin (IG) preparations designed
L Singer et al.
Pathobiology : journal of immunopathology, molecular and cellular biology, 62(1), 14-28 (1994-01-01)
The third complement component (C3) is a multifunctional glycoprotein that interacts with numerous serum proteins, cell surface receptors, and membrane-associated regulatory proteins. Deficiencies of C3 have been reported in several human kindred of different ethnic backgrounds and from different geographic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service