Skip to Content
Merck
All Photos(1)

Key Documents

RAB0630

Sigma-Aldrich

Human NPPB / Natriuretic Peptides B ELISA Kit

for serum, plasma, cell culture supernatants and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type urine
sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 14 pg/mL
standard curve range: 15.63-1000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NPPB(4879)

General description

The NPPB ELISA kit provides for the Quantitative measurement of Natriuretic Peptides B in Cell Culture Supernatants, Plasma and Serum.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunlian Zhong et al.
Oncotarget, 8(33), 54187-54198 (2017-09-15)
Myocarditis is a major cause of sudden, unexpected death in young people. However, it is still one of the most challenging diseases to treat in cardiology. In the present study, we showed that both expression level and activity of PKC-α
Markus Stenemo et al.
European journal of heart failure, 20(1), 55-62 (2017-10-03)
To identify novel risk markers for incident heart failure using proteomic profiling of 80 proteins previously associated with cardiovascular pathology. Proteomic profiling (proximity extension assay) was performed in two community-based prospective cohorts of elderly individuals without heart failure at baseline:
Emer R McGrath et al.
Journal of the American Heart Association, 9(19), e014659-e014659 (2020-09-15)
Background GDF15 (growth differentiation factor 15) and NT-proBNP (N-terminal pro-B-type natriuretic peptide) may offer promise as biomarkers for cognitive outcomes, including dementia. We determined the association of these biomarkers with cognitive outcomes in a community-based cohort. Methods and Results Plasma
Tone B Enger et al.
Journal of cardiothoracic and vascular anesthesia, 31(3), 837-846 (2016-12-28)
To investigate whether a multimarker strategy combining preoperative biomarkers representing distinct pathophysiologic pathways enhances preoperative risk assessment of acute kidney injury after cardiac surgery (CSA-AKI) and increases knowledge of underlying pathogenesis. Prospective, cohort study. Single-center tertiary referral hospital. The study
Serum Endocan Level as a Predictive Marker for Hemodynamically Significant Patent Ductus Arteriosus in Very Low Birth Weight İnfants.
Halit Halil et al.
American journal of perinatology, 34(13), 1312-1317 (2017-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service