Merck
  • Effect of PKC inhibitor on experimental autoimmune myocarditis in Lewis rats.
Effect of PKC inhibitor on experimental autoimmune myocarditis in Lewis rats.
Oncotarget (2017-09-15)
Chunlian Zhong, Yang Wu, He Chang, Chunxiao Liu, Li Zhou, Jun Zou, Zhi Qi
ABSTRACT
Myocarditis is a major cause of sudden, unexpected death in young people. However, it is still one of the most challenging diseases to treat in cardiology. In the present study, we showed that both expression level and activity of PKC-α were up-regulated in the rat heart of experimental autoimmune myocarditis (EAM). Intraperitoneal administration of PKC inhibitor (Ro-32-0432) at the end of the most severe inflammation period of EAM still significantly reduced the EAM induced expression of failure biomarkers. Furthermore, Ro-32-0432 reduced the ratio of Bax/Bcl-2 and suppressed the expression of cleaved caspase-3, both of which were increased in the heart of the EAM rats, suggesting an anti-apoptotic role of Ro-32-0432. Besides, Ro-32-0432 suppressed EAM-induced cardiac fibrosis and release of pro-inflammatory cytokines IL-1β and IL-17. These results suggest that inhibition of PKC may serve as a potential therapeutic strategy for the treatment of myocarditis.
Sigma-Aldrich
Human NPPB / Natriuretic Peptides B ELISA Kit, for serum, plasma, cell culture supernatants and urine