Skip to Content
Merck
All Photos(2)

Key Documents

RAB0429

Sigma-Aldrich

Human PAI-1 ELISA Kit

for serum, plasma, cell culture supernatant.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 35 pg/mL
standard curve range: 0.102-25 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SERPINE1(5054)

General description

The Human PAI-1 (plasminogen activator inhibitor-1) ELISA (enzyme-linked immunosorbent assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human PAI-1 in serum, plasma, cell culture supernatants. PAI-1 is 45kDa serine proteinase inhibitor. It inhibits plasmogen activator and affects the fibrinolytic process, as a result of which it leads to thrombosis. Reduced fibrinolysis is observed in ischemic heart disease. While elevated PAI-1 levels are noticed in myocardial infarction. PAI-1 is associated with various physiological processes including wound healing, sepsis, insulin resistance, obesity, atherosclerosis, tumor angiogenesis, bone remodeling, asthma, rheumatoid arthritis, glomerulonephritis and chronic stress.

Immunogen

Recombinant Human PAI-I

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pleiotropic functions of plasminogen activator inhibitor-1
Lijnen, HR
Journal of Thrombosis and Haemostasis, 3(1), 35-45 (2005)
María Paniagua-Sancho et al.
American journal of nephrology, 52(9), 714-724 (2021-09-15)
Acute kidney injury (AKI) is a threatening, multiaetiological syndrome encompassing a variety of forms and damage patterns. AKI lacks sufficiently specific diagnostic tools to evaluate the distinct combination of pathophysiological events underlying each case, which limits personalized and optimized handling.
Jessica Perugini et al.
Scientific reports, 12(1), 8331-8331 (2022-05-19)
To establish whether obesity involves activation of endogenous ciliary neurotrophic factor (CNTF) signalling, we evaluated its plasma levels in patients with obesity and correlated its values with the major clinical and haematological indices of obesity, insulin resistance and systemic inflammation.
Brain Edema XI: Proceedings of the 11th International Symposium, 277-277 (1999)
Gui Yang et al.
The Journal of allergy and clinical immunology, 133(6), 1702-1708 (2014-04-05)
The functions of regulatory T (Treg) cells are important in immunity, and the regulatory mechanisms of Treg cell activities are not fully understood yet. We sought to investigate the role of insulin-like growth factor (IGF) 2 in the upregulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service