RAB0087
Human CD40L ELISA Kit
for serum, plasma, cell culture supernatant and urine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type plasma
sample type cell culture supernatant(s)
sample type serum
sample type urine
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 8.23-6000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... CD40LG(959)
General description
The Human CD40L ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CD40L in serum, plasma (EDTA or heparin), cell culture supernatants and urine.
Immunogen
Recombinant Human CD40 Ligand
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Thrombosis research, 134(4), 832-836 (2014-08-16)
Serum soluble CD40 Ligand (sCD40L) levels, which exhibit prothrombotic and proinflammatory properties, have not been studied in patients with traumatic brain injury (TBI). Thus, the objective of this study was to determine whether serum sCD40L levels are associated with severity
The Journal of clinical endocrinology and metabolism, 99(6), E1050-E1054 (2014-03-19)
Higher serum TSH levels, both within the reference range and in those with subclinical hypothyroidism (SCH), have been associated with increased risk of atherosclerosis and cardiovascular (CV) events in a number of cross-sectional and longitudinal studies. Our objective was to
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 32(1), 151-158 (2013-10-12)
In this study, analysis and comprehensive comparison of neurogenic differentiation capacity of human bursal tissue-derived-stem cells (hBT-SCs) was aimed with human bone marrow derived mesenchymal stem cells (hBM-MSCs). hBT-SCs was isolated from subacromial bursa tissue (n = 3) by collagen
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service