Skip to Content
Merck
All Photos(2)

Key Documents

RAB0258

Sigma-Aldrich

Rat IL-13 ELISA Kit

for cell and tissue lysates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell lysate
sample type tissue lysate

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 20 pg/mL
standard curve range: 27.43-20000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

The Rat IL-13 ELSA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Rat IL-13 cell lysate and tissue lysate.

Immunogen

Recombinant Rat IL-13

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABDIL6ELISA 5X Sample Diluent Buffer (Item D2)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABLYSIS12X Cell Lysis Buffer (Item J)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arvind Chopra et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(2), 319-326 (2014-02-08)
To evaluate whether chloroquine (CQ) is more effective than meloxicam for treating early musculoskeletal pain and arthritis following acute chikungunya (CHIK) virus infection. During the 2006 CHIK epidemic, 509 rural community cases of acute CHIK virus infection were identified in
Tae Young Jang et al.
International immunopharmacology, 22(2), 444-450 (2014-08-12)
To evaluate the antiallergic effects of oral benzaldehyde in a murine model of allergic asthma and rhinitis, we divided 20 female BALB/c mice aged 8-10 weeks into nonallergic (intraperitoneally sensitized and intranasally challenged to normal saline), allergic (intraperitoneally sensitized and
Christian Schwartz et al.
Journal of immunology (Baltimore, Md. : 1950), 193(7), 3590-3599 (2014-08-31)
Schistosomiasis is a severe and chronic disease caused by the parasitic trematode Schistosoma mansoni after deposition of eggs in the liver and intestines. The immune response to S. mansoni eggs is characterized by increased Th2 cells, eosinophilia, and high serum
Eun-Hyeong Shim et al.
The Journal of pharmacy and pharmacology, 66(9), 1303-1316 (2014-06-26)
We aimed to investigate the inhibitory efficacy of Solanum tuberosum L. var. Vitelotte (SV) extract on atopic dermatitis (AD)-like skin lesions induced by the topical application of 2,4-dinitrochlorobenzene in NC/Nga mice. SV extract was administered orally to NC/Nga mice at the
Smanla Tundup et al.
Infection and immunity, 82(8), 3240-3251 (2014-05-29)
Antigen-presenting cell (APC) plasticity is critical for controlling inflammation in metabolic diseases and infections. The roles that pattern recognition receptors (PRRs) play in regulating APC phenotypes are just now being defined. We evaluated the expression of PRRs on APCs in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service