Skip to Content
Merck
All Photos(2)

Documents

RAB0302

Sigma-Aldrich

Rat IL-4 ELISA Kit

for cell and tissue lysates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell lysate
sample type tissue lysate

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.5 pg/mL
standard curve range: 0.66-160 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Il4(287287)

General description

The Rat IL-4 ELSA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Rat IL-4 cell lysate and tissue lysate.

Immunogen

Recombinant Rat IL-4

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABDIL6ELISA 5X Sample Diluent Buffer (Item D2)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABLYSIS12X Cell Lysis Buffer (Item J)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed El-Behi et al.
European journal of immunology, 44(10), 3003-3014 (2014-07-30)
IL-17-secreting CD8(+) T cells (Tc17 cells) have been implicated in immunity to infections, cancer, and autoimmune diseases. Thus far, studies on Tc17 cells have primarily investigated their development from naïve precursors, while the biology of committed Tc17 cells has been
Fen He et al.
Journal of neuroimmunology, 271(1-2), 1-7 (2014-04-10)
Patients received cranial-irradiation can be affected with cognitive deficits and decreasing hippocampal neurogenesis. In this work, we characterized the cognitive ability and immune-induced neurogenesis of the pre- and post-treated cranial-irradiated rats with Glatiramer acetate (GA), known as a weak CNS
Marwa Abdallah et al.
Experimental dermatology, 23(10), 742-747 (2014-08-12)
Not only macrophages, T-helper (Th)1 and Th2, but also CD4(+) CD25(high) FoxP3(+) regulatory T cells (T-regs) are involved in immune response to Mycobacterium leprae. We aimed to evaluate serum interleukin (IL)-1β and IL-12p70 (macrophage cytokines), interferon-γ (IFN-γ) (Th1 cytokine), IL-4
Bahaa Kenawy Abuel-Hussien Abdel-Salam
Inflammation, 37(5), 1616-1622 (2014-04-25)
The anti-inflammatory cytokines (interleukin (IL)-4 and IL-10) and the pro-inflammatory cytokines (IL-1β, IL-6, and tumor necroses factor-alpha (TNF-α)) have important functions in wound healing. Thus, the aim of this study was to determine whether dietary supplementation with whey protein could
Jing-Ge Zhang et al.
International immunopharmacology, 20(2), 307-315 (2014-04-08)
Cholecystokinin octapeptide (CCK-8), an immunomodulatory peptide, can promote or suppress the development or function of specific CD4(+) T cell subsets by regulating antigen-presenting cell functions. In the current study, we investigated whether CCK-8 exerts a direct effect on T cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service