Skip to Content
Merck
All Photos(2)

Key Documents

RAB0224

Sigma-Aldrich

Mouse IFN-γ ELISA Kit

for serum, plasma, cell culture supernatant

Synonym(s):

IFN-gamma, Interferon gamma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 5 pg/mL
standard curve range: 2.74-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Ifng(15978)

General description

The Mouse IFN-γ ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse IFN-γ in serum, plasma and cell culture supernatants.

Immunogen

Recombinant Mouse IFN-γ

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Mouse IFN-γ ELISA Kit has been used to measure serum proinflammatory cytokine levels in a renal ischemia/reperfusion injury model.[1]

Biochem/physiol Actions

Interferon γ (IFN-γ) regulates cell differentiation and inflammatory responses. It stimulates the expression of apoptotic mediators. IFN-γ is associated with rheumatoid arthritis (RA). It reduces the differentiation of T helper (Th) 17 cells and osteoclasts.[2] IFN-γ has immunomodulatory and antiviral activity. It is associated with autoimmune pathology. IFN-γ provides protection against tumor development.[3]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interferon-gamma: an overview of signals, mechanisms and functions
Schroder K, et al.
Journal of Leukocyte Biology, 75(2), 163-189 (2004)
Interferon-gamma regulates inflammatory cell death by targeting necroptosis in experimental autoimmune arthritis
Lee SH, et al.
Scientific Reports, 7(1), 10133-10133 (2017)
Qianqian Chen et al.
Journal of photochemistry and photobiology. B, Biology, 216, 112152-112152 (2021-02-21)
Emerging evidence suggests that the gut microbiota plays an important role in the pathological progression of Alzheimer's disease (AD). Photobiomodulation (PBM) therapy is believed to have a positive regulatory effect on the imbalance of certain body functions, including inflammation, immunity
Qifa Ye et al.
Experimental and therapeutic medicine, 11(4), 1499-1505 (2016-04-14)
Gynostemma pentaphyllum is a traditional Chinese medicine reported to possess a wide range of health benefits. As the major component of G. pentaphyllum, gypenoside (GP) displays various anti-inflammatory and anti-oxidant properties. However, it is unclear whether GP can protect against
Gypenoside attenuates renal ischemia/reperfusion injury in mice by inhibition of ERK signaling.
Ye Q, et. al.
Experimental and Therapeutic Medicine, 11(4), 1499-1505 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service