Skip to Content
Merck
All Photos(4)

Documents

T7539

Sigma-Aldrich

Tumor Necrosis Factor-α from mouse

TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture

Synonym(s):

mTNF-α, TNF-α

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

mouse

Quality Level

recombinant

expressed in E. coli

Assay

≥95%

form

powder

potency

0.01-0.5 ng/mL EC50

mol wt

17 kDa

packaging

pkg of 10 μg
pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg protein endotoxin

color

white

solubility

water: soluble

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Tnf(21926)

Application

Tumor Necrosis Factor-α from mouse, a potent proinflammatory cytokine has been used:
  • to activate MS1 endothelium cells
  • to induce lipolysis in adipocytes
  • to study its effect on liganded and unliganded glucocorticoid receptor (GR) in interleukin (IL)-6 and IL-8 gene regulation

Biochem/physiol Actions

Tumor necrosis factor-α (TNF-α), also known as cachectin, is expressed as a 26 kDa membrane bound protein and is then cleaved by TNF-α converting enzyme (TACE) to release the soluble 17 kDa monomer, which forms homotrimers in circulation. TNF-α plays roles in antitumor activity, immune modulation, inflammation, anorexia, cachexia, septic shock, viral replication and hematopoiesis. TNF-α is expressed by a great variety of cells, with numerous inductive and suppressive agents. Primarily, TNF-α is produced by macrophages in response to immunological challenges such as bacteria (lipopolysaccharides), viruses, parasites, mitogens and other cytokines. TNF-α is cytotoxic for many transformed cells (its namesake activity) but in normal diploid cells, it can stimulate proliferation (fibroblasts), differentiation (myeloid cells) or activation (neutrophils). TNF-α also shows antiviral effects against both DNA and RNA viruses and it induces production of several other cytokines. Although TNF-α is used in clinical trials as an antitumor agent, Sigma′s cytokine, growth factor and hormone products are for research only. TNF-α and the related molecule TNF-β (LT-α) share close structural homology with 28% amino acid sequence identity and both activate the same TNF receptors, TNFR1 and TNFR2. Mouse and human TNF-α share 79% amino acid sequence identity. Unlike human TNF-α, the mouse form is glycosylated.

Unit Definition

One unit is the amount required to induce half-maximal cytolysis of L929 cells.

Physical form

Lyophilized from 10 mM sodium phosphate, pH 7.5

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Relative contribution of adipose triglyceride lipase and hormone-sensitive lipase to tumor necrosis factor-$\alpha$ (TNF-$\alpha$)-induced lipolysis in adipocytes
Yang X, et al.
The Journal of Biological Chemistry, 286(47), 40477-40485 (2011)
Glucocorticoid-independent repression of tumor necrosis factor (TNF) $\alpha$-stimulated interleukin (IL)-6 expression by the glucocorticoid receptor a potential mechanism for protection against an excessive inflammatory response
Verhoog NJD, et al.
The Journal of Biological Chemistry, 286(22), 19297-19310 (2011)
SDF-1--CXCR4 differentially regulates autoimmune diabetogenic T cell adhesion through ROBO1--SLIT2 interactions in mice
Glawe JD, et al.
Diabetologia, 56(10), 2222-2230 (2013)
Fabrícia Lima Fontes et al.
BMC medical genetics, 16, 70-70 (2015-09-01)
Bacterial meningitis (BM) is an infectious disease that results in high mortality and morbidity. Despite efficacious antibiotic therapy, neurological sequelae are often observed in patients after disease. Currently, the main challenge in BM treatment is to develop adjuvant therapies that
Jing Deng et al.
Molecular and cellular biology, 24(23), 10161-10168 (2004-11-16)
Numerous stressful conditions activate kinases that phosphorylate the alpha subunit of translation initiation factor 2 (eIF2alpha), thus attenuating mRNA translation and activating a gene expression program known as the integrated stress response. It has been noted that conditions associated with

Articles

Lipid Induced Insulin Resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service