Skip to Content
Merck
All Photos(3)

Key Documents

R9379

Sigma-Aldrich

Rhodamine B isothiocyanate–Dextran

average mol wt ~70,000

Synonym(s):

RITC-Dextran

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171501
NACRES:
NA.56
Pricing and availability is not currently available.

mol wt

average mol wt ~70,000

Quality Level

extent of labeling

0.002-0.015 mol RITC per mol glucose

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Rhodamine B is a thiol-reactive and photostable fluorescent dye.[1]

Application

Dextran labeled with rhodamine B isothiocyanate for possible use in perfusion studies in animals.
Rhodamine B isothiocyanate-Dextran has been used:
  • to label A, CI-M6PR-GFP, Lamp1-CFP-expressing wild type mouse embryonic fibroblast (MEF) cells[2]
  • in endosome/phagosome fusion assay to determine the rate of fusion of endosomes[3]
  • as a component in poration medium to monitor Hela cell uptake[4]

Biochem/physiol Actions

Rhodamine B stains the tissue in red-violet color and emits a bright red color when fluoresced. It is highly preferred for marking insects.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiuqing Gong et al.
Lab on a chip, 10(19), 2622-2627 (2010-08-07)
We report a simple, low-cost and detachable microfluidic chip incorporating easily accessible paper, glass slides or other polymer films as the chip materials along with adhesive wax as the recycling bonding material. We use a laser to cut through the
Sidi A Bencherif et al.
Biomacromolecules, 10(9), 2499-2507 (2009-08-29)
A nanostructured hyaluronic acid (HA) hydrogel was prepared by a combination of atom transfer radical polymerization (ATRP) and Michael-type addition reaction. Biodegradable POEO(300)MA-co-PHEMA nanogels with pendent hydroxy groups were prepared by activators generated by electron transfer ATRP in cyclohexane inverse
Shikha Prasad et al.
Redox biology, 12, 58-69 (2017-02-18)
Cigarette smoking (CS) is associated with vascular endothelial dysfunction in a causative way primarily related to the TS content of reactive oxygen species (ROS), nicotine, and inflammation. TS promotes glucose intolerance and increases the risk of developing type-2 diabetes mellitus
Atg5 regulates late endosome and lysosome biogenesis
Peng JY, et al.
Science China: Life Sciences, 57(1), 59-68 (2014)
Mohammad A Kaisar et al.
Redox biology, 13, 353-362 (2017-06-25)
Recently published in vitro and in vivo findings strongly suggest that BBB impairment and increased risk for stroke by tobacco smoke (TS) closely resemble that of type-2 diabetes (2DM) and develop largely in response to common key modulators such oxidative

Questions

1–2 of 2 Questions  
  1. What is the solubility of product R9379, Rhodamine B isothiocyanate–Dextran?

    1 answer
    1. The product is soluble in water at 10mg/mL.

      Helpful?

  2. detection wavelength 알려주세요. 이 제품 production information sheet가 없네요.

    1 answer
    1. For R9379, Rhodamine B isothiocyanate-Dextran, the RITC has excitation and emission wavelengths reported at λex 553 nm; λem 627 nm in methanol.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service