Skip to Content
Merck
All Photos(1)

Documents

52194

Sigma-Aldrich

Tetramethylrhodamine isothiocyanate–Dextran

average mol wt 500,000

Synonym(s):

TRITC Dextran, TRITC-Dextran 500, TRITC-Dextran

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
NACRES:
NA.32

biological source

synthetic

form

powder or crystals

mol wt

average mol wt 500,000

extent of labeling

0.001-0.008 mol/mol TRITC glucose

fluorescence

λex 550 nm; λem 575 nm±5 nm in 0.1 M Tris pH 8.0

application(s)

cell analysis

Looking for similar products? Visit Product Comparison Guide

General description

Tetramethylrhodamine isothiocyanate–Dextran, also known as TRITC-Dextran or TRITC-Dextran 500, is a fluorescent dye and a novel drug. The fluorescent activity of TRITC-Dextran is detected at 555nm wavelength. The TRITC-Dextran molecules are released in the cells through passive diffusion due to a concentration gradient.

Application

Tetramethylrhodamine isothiocyanate–Dextran (TRITC-Dextran 500) is use as a fluorescent probe to study cell processes such as cell permeability, phagocytosis and endocytosis and to study mechanisms of biomolecular delivery.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thea van der Wijk et al.
The Journal of biological chemistry, 278(41), 40020-40025 (2003-07-23)
Osmotic swelling of Intestine 407 cells leads to an immediate increase in cell surface membrane area as determined using the fluorescent membrane dye FM 1-43. In addition, as measured by tetramethylrhodamine isothiocyanate (TRITC)-dextran uptake, a robust (>100-fold) increase in the
Margaret Clarke et al.
Journal of cell science, 115(Pt 14), 2893-2905 (2002-06-26)
The vacuolar H(+)-ATPase (V-ATPase) is a multi-subunit enzyme that plays important roles in eukaryotic cells. In Dictyostelium, it is found primarily in membranes of the contractile vacuole complex, where it energizes fluid accumulation by this osmoregulatory organelle and also in
Alexa Patzelt et al.
Journal of controlled release : official journal of the Controlled Release Society, 246, 174-182 (2016-09-20)
More and more investigations confirm that nanoparticles are incapable of overcoming the intact skin barrier in vivo. Do nanoparticles still have a future in dermal drug delivery? Unlike many other topically applied substances, nanoparticles have not been reported to utilize
Matthias Osswald et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 22(24), 6078-6087 (2016-08-16)
The role of blood-brain barrier (BBB) integrity for brain tumor biology and therapy is a matter of debate. We developed a new experimental approach using in vivo two-photon imaging of mouse brain metastases originating from a melanoma cell line to
Ngo Bich Nga Nathalie Tran et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 116, 12-16 (2016-10-25)
Hair follicle research is currently focused on the development of drug-loaded nanocarriers for the targeting of follicular structures in the treatment of skin and hair follicle-related disorders. In the present study, a dual-label nanocarrier system was implemented in which FITC-labeled

Articles

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service