Skip to Content
Merck
All Photos(2)

Documents

R0378

Sigma-Aldrich

Reactive Red 120

Powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C44H24Cl2N14O20S6Na6
CAS Number:
Molecular Weight:
1469.98
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Reactive Red 120,

form

powder

composition

Dye content, ≥50%

solubility

ethanol/water: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1c(\N=N\c2ccccc2S(O)(=O)=O)c(cc3cc(cc(Nc4nc(Cl)nc(Nc5ccc(Nc6nc(Cl)nc(Nc7cc(cc8cc(c(\N=N\c9ccccc9S(O)(=O)=O)c(O)c78)S(O)(=O)=O)S(O)(=O)=O)n6)cc5)n4)c13)S(O)(=O)=O)S(O)(=O)=O

InChI

1S/C44H30Cl2N14O20S6/c45-39-51-41(55-43(53-39)49-27-17-23(81(63,64)65)13-19-15-31(85(75,76)77)35(37(61)33(19)27)59-57-25-5-1-3-7-29(25)83(69,70)71)47-21-9-11-22(12-10-21)48-42-52-40(46)54-44(56-42)50-28-18-24(82(66,67)68)14-20-16-32(86(78,79)80)36(38(62)34(20)28)60-58-26-6-2-4-8-30(26)84(72,73)74/h1-18,61-62H,(H,63,64,65)(H,66,67,68)(H,69,70,71)(H,72,73,74)(H,75,76,77)(H,78,79,80)(H2,47,49,51,53,55)(H2,48,50,52,54,56)/b59-57+,60-58+

InChI key

OVSNDJXCFPSPDZ-YGZNCWGHSA-N

General description

Reactive Red 120 is a triazine-based azo dye.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yahya S Al-Degs et al.
Analytical and bioanalytical chemistry, 393(3), 1055-1062 (2008-11-13)
Selective polymeric extractants were prepared for preconcentration of Cibacron reactive red dye, a dye that is often applied with Cibacron reactive blue and Cibacron reactive yellow for dyeing of fabrics. The best extractant was fabricated (in chloroform) using methacrylic acid
Feifang Zhang et al.
Chemosphere, 67(4), 712-717 (2006-12-26)
In this study, an aqueous solution of purified, hydrolyzed C.I. Reactive Red 120 (RR 120, Color Index), was selected as a model to investigate the degradation pathways and to obtain additional information on the reaction intermediate formation. The dye was
Natali F Cardoso et al.
Journal of hazardous materials, 241-242, 146-153 (2012-10-09)
Spirulina platensis microalgae (SP) and commercial activated carbon (AC) were compared as adsorbents to remove Reactive Red 120 (RR-120) textile dye from aqueous effluents. The batch adsorption system was evaluated in relation to the initial pH, contact time, initial dye
Neng-Chou Shang et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 41(10), 2299-2310 (2006-10-05)
This study investigates the ozonation of Reactive Red 120 and Acid Red 299 dyes in the synthesized solution and textile wastewater by using a rotating packed bed. The decomposition rate of Reactive Red 120 and Acid Red 299 dyes via
Inhibition of the sarcoplasmic reticulum Ca2+-ATPase by Reactive Red 120.
Coll RJ and Murphy AJ
Biochimica et Biophysica Acta (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service