Skip to Content
Merck
All Photos(2)

Key Documents

B1064

Sigma-Aldrich

Cibacron Blue 3G-A

Dye content ≥55 %, Powder

Synonym(s):

Cibacron Blue; 1-amino-4-[4-[[4-chloro-6-(2-sulfoanilino)-1,3,5-triazin-2-yl]amino]-3-sulfoanilino]-9,10-dioxoanthracene-2-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H20ClN7O11S3
CAS Number:
Molecular Weight:
774.16
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

Product Name

Cibacron Blue 3G-A, Dye content ≥55 %

form

powder

composition

Dye content, ≥55%

color

dark blue

solubility

H2O: 10 mg/mL, blue

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Nc1c(cc(Nc2ccc(Nc3nc(Cl)nc(Nc4ccccc4S(O)(=O)=O)n3)c(c2)S(O)(=O)=O)c5C(=O)c6ccccc6C(=O)c15)S(O)(=O)=O

InChI

1S/C29H20ClN7O11S3/c30-27-35-28(33-16-7-3-4-8-19(16)49(40,41)42)37-29(36-27)34-17-10-9-13(11-20(17)50(43,44)45)32-18-12-21(51(46,47)48)24(31)23-22(18)25(38)14-5-1-2-6-15(14)26(23)39/h1-12,32H,31H2,(H,40,41,42)(H,43,44,45)(H,46,47,48)(H2,33,34,35,36,37)

InChI key

YKCWQPZFAFZLBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

When immobilized using an insoluble porous support matrix such as agarose, Cibacron Blue 3GA is used for affinity chromatography purification of enzymes. This affinity has been attributed to a structural similarity between the dye and the natural ligands for proteins with cofactor binding domains.

Biochem/physiol Actions

Cibacron Blue 3GA is an anionic anthraquinone dye. It acts as a P2-purinoceptor antagonist and inhibits stimulus-evoked glutamate release by rat brain cortical tissue. Cibacron Blue 3GA inhibits OXA-1 and OXA-2 β-lactamases and was used to observe the binding of ligands to OXA-1 β-lactamase by monitoring tryptophan fluorescence.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W O Smith et al.
Plant physiology, 68(2), 443-446 (1981-08-01)
The binding of phytochrome to Cibacron Blue 3GA was utilized to develop a new affinity purification procedure for phytochrome. Brushite-purified phytochrome from rye (Secale cereale c.v. Cougar) was bound to agarose-immobilized blue dye in 0.1 molar potassium phosphate (pH 7.8)
S T Thompson et al.
Proceedings of the National Academy of Sciences of the United States of America, 72(2), 669-672 (1975-02-01)
A procedure is described to utilize blue dextran-Sepharose as an affinity chromatographic column specific for the super-secondary structure called the dinucleotide fold, which forms the binding sites for substrates and effectors on a wide range of proteins. The procedure can
G C Bennett et al.
British journal of pharmacology, 131(3), 617-623 (2000-10-04)
Evidence has previously been presented that P1 receptors for adenosine, and P2 receptors for nucleotides such as ATP, regulate stimulus-evoked release of biogenic amines from nerve terminals in the brain. Here we investigated whether adenosine and nucleotides exert presynaptic control
I von Kügelgen et al.
British journal of pharmacology, 113(3), 815-822 (1994-11-01)
1. Some postganglionic sympathetic axons possess P2Y-like P2-purinoceptors which, when activated, decrease the release of noradrenaline. We examined the question of whether such receptors also occur at the noradrenergic axons in the rat brain cortex. Slices of the brain cortex
C Monaghan et al.
The Biochemical journal, 205(2), 413-417 (1982-08-01)
Although beta-lactamases do not require any nucleotide co-substrates, the OXA-2 type is inhibited competitively by Cibacron Blue 3GA, and by other anthraquinone dyes, including some simpler compounds with no side chain. The enzyme causes a red shift in the spectrum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service