Skip to Content
Merck
All Photos(1)

Documents

337714

Sigma-Aldrich

Hydroquinine

98%

Synonym(s):

(8a,9R)-10,11-Dihydro-6′-methoxycinchonan-9-ol, Dihydroquinine, NSC 41799

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26N2O2
CAS Number:
Molecular Weight:
326.43
Beilstein:
91444
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

optical activity

[α]25/D −148°, c = 1 in ethanol

mp

168-176 °C (lit.)

SMILES string

CC[C@H]1CN2CCC1C[C@@H]2[C@H](O)c3ccnc4ccc(OC)cc34

InChI

1S/C20H26N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h4-6,8,11,13-14,19-20,23H,3,7,9-10,12H2,1-2H3/t13-,14-,19-,20+/m0/s1

InChI key

LJOQGZACKSYWCH-WZBLMQSHSA-N

Gene Information

human ... CYP2D6(1565)

Looking for similar products? Visit Product Comparison Guide

General description

Hydroquinine is a cinchona alkaloid.

Application

Hydroquinine may be used in the preparation of its derivatives such as C9 epihydroquinine, 9-acetoxy-10,11-dihydroquinine and 10,11-dihydroquinine monohydrochloride. It may also be used as a catalyst for the methylhydrazine promoted asymmetric α-hydroxylation of β-keto esters in the presence of molecular oxygen as oxidant to form enantioenriched α-hydroxy β-dicarbonyl compounds.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ion-pair formation of hydroquinine by chromatography.
Marchais S, et al.
Analytica Chimica Acta, 426(1), 85-93 (2001)
Li-Ping Zhang et al.
Analytical and bioanalytical chemistry, 412(26), 7321-7332 (2020-08-14)
A novel soluble molecularly imprinted polymer (SMIP) without chemical cross-linker was successfully synthesized. The quinine (QN), which the structure was similar to the template, was chosen as the immobile template to improve the affinity of MIP. 4-Methyl phenyl dicyclohexyl ethylene
Methylhydrazine-induced enantioselective a-hydroxylation of ?-keto esters with molecular oxygen catalyzed by hydroquinine.
Wang Y, et al.
Tetrahedron, 71(1), 85-90 (2015)
Fiorenzo Gaita et al.
Journal of the American College of Cardiology, 43(8), 1494-1499 (2004-04-20)
The purpose of this study was to evaluate the efficacy of various antiarrhythmic drugs at prolonging the QT interval into the normal range and preventing ventricular arrhythmias in patients with short QT syndrome. Short QT syndrome is a recently described
Ad Kane et al.
Annales de cardiologie et d'angeiologie, 61(4), 292-295 (2011-06-15)
A 45-year-old man was hospitalized for syncope due to fascicular ventricular tachycardia degenerating into ventricular fibrillation (VF). The electrocardiogram showed an early repolarization syndrome. The arrhythmia was repetitive and disappeared after oral hydroquinidine. An implantable cardioverter-defibrillator (ICD) was implanted; subsequently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service