Skip to Content
Merck
All Photos(3)

Documents

PZ0014

Sigma-Aldrich

Linezolid

≥98% (HPLC), powder, bacterial mRNA translation inhibitor

Synonym(s):

N-[[(5S)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]acetamide, PNU-100766, U-100766

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20FN3O4
CAS Number:
Molecular Weight:
337.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Linezolid, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

room temp

SMILES string

CC(=O)NC[C@H]1CN(C(=O)O1)c2ccc(N3CCOCC3)c(F)c2

InChI

1S/C16H20FN3O4/c1-11(21)18-9-13-10-20(16(22)24-13)12-2-3-15(14(17)8-12)19-4-6-23-7-5-19/h2-3,8,13H,4-7,9-10H2,1H3,(H,18,21)/t13-/m0/s1

InChI key

TYZROVQLWOKYKF-ZDUSSCGKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Linezolid exhibits its activity against several Gram-positive bacteria including penicillin-resistant, methicillin-resistant, and vancomycin- resistant. Linezolid can decrease the rate of translation reaction by inhibiting the formation of initiation complex thereby reducing the length of the newly formed peptidechain. Additionally, it may also play a role in reducing the virulence leading to reduced toxins produced by Gram-positive bacteria.

Application

Linezolid has been used in checkerboard-type drug combination MIC (minimum inhibitory concentrations) assay to determine whether ivacaftor shows positive interaction with linezolid. It has been used for the determination of minimum biofilm inhibitory concentration and for the treatment of Mycobacterium abscessus-infected Drosophila melanogaster w1118 flies.Linezolid has been used as an antibiotic to study its effects on the development of malaria-associated acute respiratory distress syndrome (MAARDS) in rat lungs. It has been used as an antibiotic to study its effects in combination with baicalein on methicillin-resistant Staphylococcus aureus strains. It has been used as an antibiotic to study its effects on planktonic growth and formation of biofilms by Enterococcus faecalis.

Biochem/physiol Actions

Linezolid is an oxazolidinone antimicrobial. It binds to a site on the bacterial 23S ribosomal RNA of the 50S subunit and prevents the formation of a functional 70S initiation complex, thus inhibiting bacterial mRNA translation. Linezolid is also a weak, reversible, nonselective inhibitor of monoamine oxidase.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Bone

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjay Chhibber et al.
PloS one, 8(2), e56022-e56022 (2013-02-19)
Staphylococcus aureus remains the predominant pathogen in diabetic foot infections and prevalence of methicillin resistant S.aureus (MRSA) strains further complicates the situation. The incidence of MRSA in infected foot ulcers is 15-30% and there is an alarming trend for its
Matthew S Dryden
The Journal of antimicrobial chemotherapy, 66 Suppl 4, iv7-iv15 (2011-05-06)
Linezolid has been widely used in the treatment of Gram-positive infections for more than a decade. It is unique amongst antibiotics active against most multiply-resistant Gram-positive bacteria in that there is an oral preparation with 100% bioavailability and an extensive
Gregory J Moran et al.
The Lancet. Infectious diseases, 14(8), 696-705 (2014-06-10)
New antibiotics are needed to treat infections caused by drug-resistant bacteria. Tedizolid is a novel oxazolidinone antibacterial drug designed to provide enhanced activity against Gram-positive pathogens. We aimed to assess the efficacy and safety of intravenous to oral tedizolid for
Giovanni Sotgiu et al.
The European respiratory journal, 40(6), 1430-1442 (2012-04-13)
Linezolid is used off-label to treat multidrug-resistant tuberculosis (MDR-TB) in absence of systematic evidence. We performed a systematic review and meta-analysis on efficacy, safety and tolerability of linezolid-containing regimes based on individual data analysis. 12 studies (11 countries from three
Bing Gu et al.
The Journal of antimicrobial chemotherapy, 68(1), 4-11 (2012-09-06)
The oxazolidinone antibiotic linezolid has demonstrated potent antimicrobial activity against Gram-positive bacterial pathogens, including methicillin-resistant staphylococci. This article systematically reviews the published literature for reports of linezolid-resistant Staphylococcus (LRS) infections to identify epidemiological, microbiological and clinical features for these infections.

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Related Content

Explore protein pathway analysis through chemical library screening, investigating protein interactions, and modulating signaling pathways with small molecules.

Explore protein pathway analysis through chemical library screening, investigating protein interactions, and modulating signaling pathways with small molecules.

Explore protein pathway analysis through chemical library screening, investigating protein interactions, and modulating signaling pathways with small molecules.

Explore protein pathway analysis through chemical library screening, investigating protein interactions, and modulating signaling pathways with small molecules.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service