Skip to Content
Merck
All Photos(4)

Key Documents

C8895

Sigma-Aldrich

Clofazimine

≥98% (TLC), powder, antimycobacterial rimonphenazine

Synonym(s):

N,5-Bis(4-chlorophenyl)-3,5-dihydro-3-(isopropylimino)phenazin-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H22Cl2N4
CAS Number:
Molecular Weight:
473.40
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Clofazimine,

form

powder

antibiotic activity spectrum

mycobacteria

Mode of action

protein synthesis | interferes

originator

Novartis

SMILES string

CC(C)\N=C1/C=C2N(c3ccc(Cl)cc3)c4ccccc4N=C2C=C1Nc5ccc(Cl)cc5

InChI

1S/C27H22Cl2N4/c1-17(2)30-24-16-27-25(15-23(24)31-20-11-7-18(28)8-12-20)32-22-5-3-4-6-26(22)33(27)21-13-9-19(29)10-14-21/h3-17,31H,1-2H3/b30-24+

InChI key

WDQPAMHFFCXSNU-BGABXYSRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Clofazimine is a riminophenazine, which is used as an antileprosy drug. It is used to treat multidrug-resistant tuberculosis. Clofazimine prevents neutrophil motility and lymphocyte transformation. It has anti-inflammatory effect, which is used to treat discoid lupus erythematosus. Clofazimine has immunosuppressive property.

Application

Clofazimine has been used:
  • for antimicrobial preparation
  • to study its accumulation on macrophages to form crystal-like drug inclusions (CLDIs)
  • to model drug-induced hepatic granulomatous inflammation
  • to study the in vivo cargo storage capacity of macrophages

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage and Stability

This product is stored at room temperature.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mode of action of clofazimine and combination therapy with benzothiazinones against Mycobacterium tuberculosis
Lechartier B and Cole ST
Antimicrobial Agents and Chemotherapy, 59(8), 4457-4463 (2015)
Macrophage-mediated clofazimine sequestration is accompanied by a shift in host energy metabolism
Trexel J, et al.
Journal of Pharmaceutical Sciences, 106(4), 1162-1174 (2017)
The Antimicrobial Drugs (2000)
Sarah Schmidt Grant et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(30), 12147-12152 (2012-07-11)
During Mycobacterium tuberculosis infection, a population of bacteria likely becomes refractory to antibiotic killing in the absence of genotypic resistance, making treatment challenging. We describe an in vitro model capable of yielding a phenotypically antibiotic-tolerant subpopulation of cells, often called
Detecting ordered small molecule drug aggregates in live macrophages: a multi-parameter microscope image data acquisition and analysis strategy
Rzeczycki P, et al.
Biomedical Optics Express, 8(2), 860-872 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service