Skip to Content
Merck
All Photos(1)

Key Documents

PPC1010

Sigma-Aldrich

Protease and Phosphatase Inhibitor Cocktail

DMSO solution, for the inhibition of serine, cysteine, acid proteases, aminopeptidase, serine/threonine protein phosphatases and L-isozymes of alkaline phosphatase

Synonym(s):

Protease and Phosphatase Inhibitor Mix, Protease and phosphatase inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.54
form:
DMSO solution
solubility:
water: soluble
storage temp.:
−20°C

Product Name

Protease and Phosphatase Inhibitor Cocktail,

form

DMSO solution

Quality Level

packaging

pkg of 1 ml, 5 ml ×

solubility

water: soluble

shipped in

wet ice

storage temp.

−20°C

General description

Protease and Phosphatase Inhibitor Cocktail is designed to increase the yield of intact and unmodified proteins in cell extracts. Protease and Phosphatase Inhibitor cocktail mixture inhibits serine, cysteine, and acid proteases, as well as aminopeptidase. It also inhibits serine/threonine protein phosphatases and L-isozymes of alkaline phosphatase. The cocktail has been tested on extracts from various animal tissues such as the human placenta and bovine liver and on extracts of A431, CHO, and U937 cells. Protease and Phosphatase Inhibitor Cocktail is supplied as a ready-to-use solution in DMSO.

Application

Protease and Phosphatase Inhibitor Cocktail has been used as a supplement in radioimmunoprecipitation assay (RIPA) buffer:
  • for cellular protein extraction
  • to lyse purified CD4+ T cells
  • to prepare protein extracts from placental tissues and cells

Protease and Phosphatase Inhibitor Cocktail has been used in protein isolation buffers.

Preparation Note

The recommended dilution of the cocktail in the biological extract is 1 ml of the cocktail per 500 mg of protein extracted from tissue or cells. In many cases, the cocktail can be used at a final concentration of 1% (v/v), 1 ml of cocktail solution per 100 ml of extraction buffer.

Other Notes

This product is for R&D use only, not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TAGLN2-regulated trophoblast migration, invasion and fusion are impaired in preeclampsia
Wang H, et al.
Frontiers in Cell and Developmental Biology, 10, 810633-810633 (2022)
Beta-Arrestin1 Prevents Preeclampsia by Downregulation of Mechanosensitive AT1-B2 Receptor Heteromers
Quitterer U, et al.
Cell, S0092-8674(18), 31406-31406 (2018)
Marcela Rassi-Cruz et al.
Endocrine-related cancer, 28(1), 1-13 (2020-10-29)
Familial primary aldosteronism (PA) is rare and mostly diagnosed in early-onset hypertension (HT). However, 'sporadic' bilateral adrenal hyperplasia (BAH) is the most frequent cause of PA and remains without genetic etiology in most cases. Our aim was to investigate new
E Zoico et al.
Mechanisms of ageing and development, 192, 111374-111374 (2020-10-06)
Fibrosis has been considered as a hallmark of dysfunctional adipose tissue (AT), however the role and mechanisms of fibrosis in the age related AT dysfunction are not yet well characterized. The aim of the study was to investigate the mechanisms
LKB1-PTEN axis controls Th1 and Th17 cell differentiation via regulating mTORC1
Pandit M, et al.
Journal of Molecular Medicine, 99(8), 1139-1150 (2021)

Related Content

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service