Skip to Content
Merck
All Photos(1)

Key Documents

P9797

Sigma-Aldrich

Pyridostigmine bromide

Synonym(s):

3-(Dimethylaminocarbonyloxy)-1-methylpyridinium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13BrN2O2
CAS Number:
Molecular Weight:
261.12
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

solubility

water: 100 mg/mL, clear to very slightly hazy, colorless

originator

Valeant

SMILES string

[Br-].CN(C)C(=O)Oc1ccc[n+](C)c1

InChI

1S/C9H13N2O2.BrH/c1-10(2)9(12)13-8-5-4-6-11(3)7-8;/h4-7H,1-3H3;1H/q+1;/p-1

InChI key

VNYBTNPBYXSMOO-UHFFFAOYSA-M

Gene Information

human ... ACHE(43)

Looking for similar products? Visit Product Comparison Guide

Application

Pyridostigmine bromide has been used as a cholinesterase inhibitor in mice, in florimetric assay for acetylcholine secretion and for measuring acetylcholine release from mice atrial tissue.

Biochem/physiol Actions

Acetylcholinesterase inhibitor.
Pyridostigmine bromide inhibits acetylcholine esterase and is membrane impermeable. It is used as a preventive medicine in chemical warfare. Pyridostigmine bromide is a carbamate compound comprising quaternary amine structure. It displays low absorption at the gastrointestinal tract. Sustained release of pyridostigmine is recommended for improving its bioavailability in treating myasthenia gravis.

Features and Benefits

This compound was developed by Valeant. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cardiomyocyte-secreted acetylcholine is required for maintenance of homeostasis in the heart
Roy A, et al.
Faseb Journal, 27(12), 5072-5082 (2013)
Gulf War and health Depleted uranium, pyridostigmine bromide, sarin, and vaccines (2000)
Benefits from sustained-release pyridostigmine bromide in myasthenia gravis: results of a prospective multicenter open-label trial
Sieb JP and Kohler W
Clinical Neurology and Neurosurgery, 112(9), 781-784 (2010)
Protein binding of isofluorophate in vivo after coexposure to multiple chemicals.
Vogel JS, et al.
Environmental Health Perspectives, 110(suppl 6), 1031-1036 (2002)
Preparation and in vitro evaluation of pyridostigmine bromide microparticles
Hegazy N, et al.
International Journal of Pharmaceutics, 242(1-2), 171-174 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service