Skip to Content
Merck
All Photos(3)

Documents

A8003

Sigma-Aldrich

Allopurinol

xanthine oxidase inhibitor

Synonym(s):

1H-Pyrazolo(3,4-d)pyrimidin-4-ol, 4-Hydroxypyrazolo(3,4-d)pyrimidine, 4-Hydroxypyrazolo[3,4-d]pyrimidine, HPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N4O
CAS Number:
Molecular Weight:
136.11
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥99% (TLC)

form

powder

mp

>300 °C (lit.)

solubility

1 M NaOH: soluble 50 mg/mL, clear to very slightly hazy, colorless to faintly yellow

storage temp.

room temp

SMILES string

O=C1NC=Nc2[nH]ncc12

InChI

1S/C5H4N4O/c10-5-3-1-8-9-4(3)6-2-7-5/h1-2H,(H2,6,7,8,9,10)

InChI key

OFCNXPDARWKPPY-UHFFFAOYSA-N

Gene Information

human ... XDH(7498)

Looking for similar products? Visit Product Comparison Guide

Application

Allopurinol has been used:
  • to decrease the uric acid levels and study its effect on renal injury and inflammation in mice
  • as a xanthine oxidase inhibitor to test panton-valentine leukocidin (PVL) neutrophil extracellular traps (NETosis) dependence on different enzymes, channels, or organelles using human polymorphonuclear cells (hPMNs)
  • as a reference standard to study the inhibitory effect of olive leaf components on xanthine oxidase in vitro

Allopurinol is suitable for:
  • intravenous injection in mice for inhibition of xanthine oxidase
  • use as an inhibitor of xanthine oxidase
  • use as a positive control in xanthine oxidase inhibition assay
  • use in the preparation of University of Wisconsin solution for osmotic stabilization to enhance the density differences between islets and acinar fragments during porcine islet purification

Biochem/physiol Actions

Allopurinol is a purine analog that inhibits certain enzymes involved in purine metabolism. It is known to reduce the synthesis of uric acid in the body. Allopurinol mediated inhibition of xanthine oxidase exhibits anti-inflammatory effects on atherosclerosis, acute lung injury, and congestive heart failure.
Inhibitor of xanthine oxidase and de novo pyrimidine biosynthesis. A classical agent in treatment of hyperuricemia and gout.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Kubo et al.
The Journal of clinical investigation, 97(11), 2680-2684 (1996-06-01)
Mice with chronic granulomatous disease (X-CGD mice) generated by mutating the X-linked gene for a subunit of NADPH oxidase have been analyzed for their ability to respond to intravenous injection of purified cobra venom factor (CVF). This agent in wild-type
Andreas Haryono et al.
The Kobe journal of medical sciences, 64(3), E107-E114 (2019-01-23)
Hyperuricemia contributed to endothelial dysfunction, activation of the RAS system, increased oxidative stress and maladaptive immune system response. M1 and M2 macrophages were known to contribute to the onset of renal fibrosis. This study aimed to look at the effect
Nitrate partially inhibits lipopolysaccharide-induced inflammation by maintaining mitochondrial function.
Yang Yang et al.
The Journal of international medical research, 48(2), 300060520902605-300060520902605 (2020-02-12)
J Flemmig et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 18(7), 561-566 (2010-12-15)
In Mediterranean folk medicine Olea europaea L. leaf (Ph.Eur.) preparations are used as a common remedy for gout. In this in vitro study kinetic measurements were performed on both an 80% ethanolic (v/v) Olea europaea leaf dry extract (OLE) as
Michael P M van der Burg et al.
TheScientificWorldJournal, 3, 1154-1159 (2003-12-04)
Generally, prior to the purification of isolated pancreatic islets, the collagenase-digested tissue is incubated in the University of Wisconsin solution (UWS; approximately 320 mOsm) for osmotic stabilization to preserve or improve the density differences between islets and acinar fragments. The

Articles

Discover critical characteristics to consider when working with enzyme inhibitors, such as cell permeability, the prozone effect, and Lipinski’s rule of 5.

Discover critical characteristics to consider when working with enzyme inhibitors, such as cell permeability, the prozone effect, and Lipinski’s rule of 5.

Discover critical characteristics to consider when working with enzyme inhibitors, such as cell permeability, the prozone effect, and Lipinski’s rule of 5.

Discover critical characteristics to consider when working with enzyme inhibitors, such as cell permeability, the prozone effect, and Lipinski’s rule of 5.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service