Skip to Content
Merck
All Photos(3)

Key Documents

N2001

Sigma-Aldrich

Neostigmine bromide

≥98% (HPLC and titration), powder

Synonym(s):

3-(N,N-Dimethylcarbamoyloxy)-N,N,N,-trimethylanilinium bromide, Prostigmine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19BrN2O2
CAS Number:
Molecular Weight:
303.20
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC and titration)

form

powder

color

white

mp

175-177 °C (lit.)

solubility

H2O: 1 g/mL
ethanol: soluble

SMILES string

[Br-].CN(C)C(=O)Oc1cccc(c1)[N+](C)(C)C

InChI

1S/C12H19N2O2.BrH/c1-13(2)12(15)16-11-8-6-7-10(9-11)14(3,4)5;/h6-9H,1-5H3;1H/q+1;/p-1

InChI key

LULNWZDBKTWDGK-UHFFFAOYSA-M

Gene Information

human ... ACHE(43)

Looking for similar products? Visit Product Comparison Guide

Application

Neostigmine bromide has been used as an acetyl cholinesterase (AChE) inhibitor:
  • to control reaction specificity of acetylcholinesterase in coronal organ of Botryllus schlosseri.
  • to study its effects on abnormal contraction of pyloric circular smooth muscle strips (PCSMS).
  • to study its effects on depolarization of bag cell neurons.

Biochem/physiol Actions

Neostigmine bromide is a quaternary amine. It is involved in elongating and boosting the muscarinic and nicotinic effects of acetylcholine by inhibiting cholinesterase activity. It is used in anesthesia to reverse the neuromuscular blockade produced by competitive neuromuscular blockers.
Reversible inhibitor of acetylcholinesterase that is similar to eserine, but does not cross the blood-brain barrier.
Reversible inhibitor of acetylcholinesterase; does not cross the blood-brain barrier.

Features and Benefits

This compound is featured on the Acetylcholine Synthesis and Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modified carbon paste sensor for the potentiometric determination of neostigmine bromide in pharmaceutical formulations, human plasma and urine
Khorshid, A F and Issa, Y M
Biosensors And Bioelectronics, 51, 143-149 (2014)
Effect of aurantii fructus immaturus flavonoid on the contraction of isolated gastric smooth muscle strips in rats
Wu Z, et al.
Evidence-Based Complementary and Alternative Medicine : ECAM, 2016(2), 317-328 (2016)
Zhenyu Wu et al.
Evidence-based complementary and alternative medicine : eCAM, 2016, 5616905-5616905 (2016-07-19)
This study was designed to investigate the effect of Aurantii fructus immaturus flavonoid (AFIF) on the contraction of isolated gastric smooth muscle in rats and explore its underlying mechanisms. Isolated antral longitudinal smooth muscle strip (ALSMS) and pyloric circular smooth
Differentiation and induced sensorial alteration of the coronal organ in the asexual life of a tunicate
Manni L, et al.
Integrative and Comparative Biology, 58(2), 317-328 (2018)
A F Khorshid et al.
Biosensors & bioelectronics, 51, 143-149 (2013-08-21)
A novel, simple, rapid, selective and sensitive method for the determination of neostigmine (Ns) ion in its bulk powder, different pharmaceutical dosage forms, and biological fluids (plasma and urine) using four modified carbon paste electrodes was developed. Sensor 1 is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service